NIOSH 7401 (Issue 2)
Alkaline Dusts
This test method uses an acid-base titration technique to measure total alkalinity of alkali hydroxides, carbonates, borates, silicates, phosphates, and other basic salts, expressed as equivalents of NaOH. The method measures the total alkalinity of alkali hydroxides, carbonates, borates, silicates, phosphates, and other basic salts. This revises Methods S381 and P&CAM 241.
Analysis Type (Application Method): Acid-base titration
Link to NIOSH 7401 (Issue 2) Method
Compatible Filters:
37 mm 1.0 μm PTFE supported by a cellulose backup pad in a cassette filter holder, syringe 0.45-μm PTFE filter for mobile phase prep. Forceps to handle filter membrane.