NIOSH 7505
LEAD SULFIDE
Lead sulfite is a semiconducting material with niche uses.This method uses x-ray powder diffraction to monitor lead sulfide. This method is specific for lead sulfide among other mine dusts. This method was originally designated P&CAM 350, which it replaces. THF is extremely flammable and should be used in a fume hood.
Analysis Type (Application Method): X-Ray powder diffraction
Link to NIOSH 7505 Method
Compatible Filters:
37 mm 5 μm PVC filter with a backup pad in a 2 piece 37 mm cassette, 25 mm 0.45 μm silver membrane