NIOSH 7506
BORON CARBIDE
This method uses x-ray powder diffraction for determining respirable boron carbide among other mine dusts. This revises P&CAM 324.Boron carbide is a material used in tank armor, bulletproof vests, and engine sabotage powders. It is one of the hardest known materials.
Analysis Type (Application Method): X-Ray powder diffraction
Link to NIOSH 7506 Method
Compatible Filters:
37 mm 5 μm PVC filter supported w/backup pad. Forceps to handle filter membrane.
Materials
Related Links
Categories