NIOSH 7600
CHROMIUM, HEXAVALENT
This test method uses visible absorption spectrophotometry techniques to monitor Chromium Hexavalent. This method combines and replaces P&CAM 169, S317 and P&CAM 319; the Cr(VI) criteria document contains a method similar to P&CAM 169. All sample preparation should be performed in a hood.
Analysis Type (Application Method): Visible absorption, spectrophotometry
Link to NIOSH 7600 Method
Compatible Filters:
37 mm 5 μm PVC in polyester cassette filter holder. Forceps to handle filter membrane.