NIOSH 7602 (Issue 3)
SILICA, CRYSTALLINE by IR (KBr pellet)
This test method uses a infrared absorption spectrophotometry to detect Cristobalite and tridymite. Keep away from flames. Use personal protective equipment to prevent contact of acids with the skin. This is P&CAM 110 in a revised format.
Analysis Type (Application Method): IR
Link to NIOSH 7602 (Issue 3) Method
Compatible Filters:
37 mm 5.0 μm PVC supported with backup pad in a two-piece, 37 mm cassette filter holder (preferably, conductive) held together by tape or cellulose shrink band.