NIOSH 7605
CHROMIUM, HEXAVALENT by Ion Chromatography
This method can be used for the determination of Cr(VI) using 2% NaOH - 3%Na2CO3for extraction. All sample preparation should be performed in a hood. Chromium Hexavalent are chemical compounds that contain choromium in the +6 oxidation state. Hexavalent chromium is used in textile dyes, wood preservation, anti-corrosion products, and chromate conversion coatings.
Analysis Type (Application Method): Ion chromatography, with post column derivatization and UV detection
Link to NIOSH 7605 Method
Compatible Filters:
37 mm 5 μm PVC membrane in polystyrene cassette filter holder. Forceps to handle filter membrane.