NIOSH 7701
LEAD BY PORTABLE ULTRASONIC EXTRACTION/ASV
For this method, lead determination by ultrasonic extraction/ASV method is applicable to the on-site, field-based determination of lead in air filter samples, and also may be used for laboratory-based air filter sample preparation and analysis.
Analysis Type (Application Method): Portable anodic stripping voltammetry
Link to NIOSH 7701 Method
Compatible Filters:
37 mm 0.8 μm MCE. Forceps to handle filter membrane.
Materials
Related Links
Categories