NIOSH 7702
LEAD BY FIELD PORTABLE XRF
This is a field portable analytical method, particularly useful for the analysis of initial exposure assessment samples, or for applications where laboratory analysis is impractical. Additionally, the method is non-destructive; samples analyzed in the field can later be analyzed in a laboratory. The method is applicable to all elemental lead forms, including lead fume, and all other aerosols containing lead.
Analysis Type (Application Method): X-ray fluorescence (XRF), portable L-shell excitation
Link to NIOSH 7702 Method
Compatible Filters:
37 mm 0.8 μm MCE. Forceps to handle filter membrane.