NIOSH 7901
ARSENIC TRIOXIDE, as As
This method collects particulate arsenic compounds as well as arsenic trioxide vapor. If only total particulate arsenic is of interest, the use of the treated filter and analysis of the backup pad is not required. Arsine is not collected by this sampling method.
Analysis Type (Application Method): Atomic absorption, graphite furnace
Link to NIOSH 7901 Method
Compatible Filters:
37 mm 0.8 μm MCE in cassette filter holder. Forceps to handle filter membrane.