NIOSH 7902
FLUORIDES, aerosol and gas by ISE
This method applies to mining samples, and samples from the aluminum reduction, ceramic, glass etching, electroplating, semi-conductor, and fluorochemical industri es. If other aerosols are present, gaseous fluoride may be underestimated owing to sorption on collected particles, with concur rent overestimation of particulate/gaseous fluoride ratio.
Analysis Type (Application Method): Ion specific electrode (ISE)
Link to NIOSH 7902 Method
Compatible Filters:
37 mm 0.8 μmMCE with nucleopore and cellulose pad in a 2 piece cassette filter holder. Forceps to handle filter membrane.
Materials
Related Links
Categories