NIOSH 7907
VOLATILE ACIDS by Ion Chromatography (Hydrogen Chloride, Hydrogen Bromide, Nitric Acid)
This method is the replacement method NIOSH 7903 and allows for the collection of the inhalable fraction of particulate fluorides. Improvements with this method allows lower limits of detection and quantitation for gaseous HF due to higher sampling flow rates.
Analysis Type (Application Method): Ion Chromatography
Link to NIOSH 7907 Method
Compatible Filters:
37mm Quartz pre-filter and sampling filter, 37mm empty monitor, 13mm PTFE syringe filters, and forceps