NIOSH 9111 (Issue 1)
METHAMPHETAMINE On Wipes by Liquid Chromatography/Mass Spectrometry
This method uses a liquid chromatography and mass spectrometry-sim mode technique to detect methamphetamine. Avoid skin contact. Work should be performed in an adequate hood. NIOSH 9106 uses liquid-liquid extraction and gas chromatography/mass spectrometry (GC/MS) to measure multiple drugs. NIOSH 9109 uses solid-phase extraction and GC/MS to measure multiple drugs.
Analysis Type (Application Method): LC-MS SIM mode
Link to NIOSH 9111 (Issue 1) Method
Compatible Filters:
0.45 μm PES syringe filter such as the fast flowing 0.45 μm 33mm Millex® HP.