OSHA 105
INORGANIC ARSENIC IN WORKPLACE ATMOSPHERES
For this method, samples are collected closed-face by drawing known volumes of air through sampling devices consisting of three-piece cassettes, each containing two sulfuric acid-treated glass fiber filters separated by the ring section. Samples are analyzed by HPLC using an ultraviolet detector.
Analysis Type (Application Method): AAS, graphite furnace
Link to OSHA 105 Method
Compatible Filters:
37 mm 0.8 μm MCE in cassette filter holder. Forceps to handle filter membrane.