OSHA 145
Particulate Mercury in Workplace Atmospheres
This method describes the collection of airborne particulate mercury on 0.8-μm mixed-cellulose ester membrane (MCE) filters and the subsequent analysis using a cold vapor-atomic absorption spectrophotometer (CV-AAS). Wipe and bulk samples can also be collected and analyzed for mercury content using this method.
Analysis Type (Application Method): Cold Vapor Atomic Absorption (CVAAS)
Link to OSHA 145 Method
Compatible Filters:
37 mm 0.8 μm MCE in cassette filter holder. Forceps to handle filter membrane.