OSHA 215
Hexavalent Chromium
This test method uses ses HPLC to monitor Chromium Hexavalent. This method should be used by industrial hygienists experienced in monitoring for exposures and analysts experienced in the use of ion chromatography and the interpretation of ion chromatograms. Samples are taken in the breathing zone of workplace personnel, and analysis is performed with an ion chromatograph (IC) equipped with a UV-vis detector and a postcolumn reagent delivery system.
Analysis Type (Application Method): HPLC
Link to OSHA 215 Method
Compatible Filters:
37 mm 5 μm PVC membrane filter in polystyrene cassette filter holder. Forceps to handle filter membrane.