OSHA 39
Pentachlorophenol
This method is nonspecific and determines the total dust concentration to which a worker is exposed. It may be applied, e.g., to gravimetric determination of fibrous glass in addition to the other ACGIH particulates not otherwise regulated. Pentachlorophenol is a compound that has been used as a pesticide and a disinfectant. It helps perserve wood and cool tower water.
Analysis Type (Application Method): HPLC
Link to OSHA 39 Method
Compatible Filters:
8 mm glass fiber filter disc and 35 mm glass fiber filter. Forceps to handle filter membrane.