OSHA 81
Crotonaldehyde
A sample is collected by drawing air through an open face air Monitoring cassette containing two glass fiber filters, each of which is coated with 2,4-dinitrophenylhydrazine and phosphoric acid. The sample is extracted with acetonitrile and analyzed by HPLC using a UV detector. As an unsaturated aldehyde, crotonaldehyde is a versatile intermediate in organic synthesis. It occurs in a variety of foodstuffs (soybean oils).
Analysis Type (Application Method): HPLC-UV
Link to OSHA 81 Method
Compatible Filters:
Two glass fiber filters in a cassette. Forceps to handle filter membrane.