OSHA 95
Ethylene Thiourea
Ethylene thiourea is used as an accelerator in synthetic rubber production, a curing agent for epichlorohydrin elastomers, and component of ethylenebisdithiocarbamate fungicides. For this method, samples are collected open-face by drawing air through a four-piece sampling cassette containing two glass fiber filters. Samples are extracted with water and analyzed by HPLC using a UV detector. Ethylene thiourea is used
Analysis Type (Application Method): HPLC-UV
Link to OSHA 95 Method
Compatible Filters:
1 μm glass fiber filters in a 4 piece polystyrene cassette filter holder. Forceps to handle filter membrane.