OSHA ID-113
Sulfuric Acid in Workplace Atmospheres
This method describes the collection and analysis of airborne sulfuric acid using ion chromatography. The method is collected by a known volume of air that drawn through a mixed cellulose ester membrane filter (MCEF). Sulfuric acid has a wide range of end applications including in domestic acidic drain cleaners, as an electrolyte in lead-acid batteries and in various cleaning agents.
Analysis Type (Application Method): IC
Link to OSHA ID-113 Method
Compatible Filters:
Filter holder, 2 or 3 piece cassette, polystyrene 37 mm in diameter. MCE filter, 0.8 μm pore size, 37 mm diameter, supported by a cellulose backup padForceps to handle filter membrane.