OSHA ID-200
SULFUR DIOXIDE IN WORKPLACE ATMOSPHERES (IMPREGNATED ACTIVATED BEADED CARBON)
This method describes the sample collection and analysis of airborne sulfur dioxide (SO2). Samples are taken in the breathing zone of workplace personnel, and analysis is performed by ion chromatography (IC). Sulfur dioxide is a toxic gas that is produced as a by-product of the burning of fossil fuels contaminated with sulfur compound.
Analysis Type (Application Method): IC
Link to OSHA ID-200 Method
Compatible Filters:
Filter for particulate collection, PTFE, 0.45 μm, 25 mm. Carbon-filled polypropylene cassette, 25 mmForceps to handle filter membrane.