[Skip to Content](https://www.sigmaaldrich.com#main-content) [![MilliporeSigma](https://www.sigmaaldrich.com/static/logos/purple/millipore_sigma.svg)](https://www.sigmaaldrich.com/US/en) Products Cart0 USEN Products ProductsApplicationsServicesResourcesSupport [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/US/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/US/en)[Gas Chromatography (GC)](https://www.sigmaaldrich.com/US/en/applications/analytical-chemistry/gas-chromatography)MSTFA/MSTFA-d9 Derivatization of Amphetamine for GC/MS Detection and Identification # MSTFA/MSTFA-d9 Derivatization of Amphetamine for GC/MS Detection and Identification ![contributed-article](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/protocols/analytical-chemistry/gas-chromatography/contributed-article.jpg "contributed-article") The following was generated with the assistance of an outside source using Sigma-Aldrich products. Technical content was generated and provided by: Dieter Urbach, Bundeskriminalamt (BKA), KT 12 – Zentrale Analytik II, 65173 Wiesbaden, Germany ## Introduction Derivatization, especially silylation, is commonly used to identify and quantify both known and unknown compounds using GC-MS. It is not limited to small molecules. Even peptides can be silylated, especially when the introduction of deuterium is desired[1](https://www.sigmaaldrich.com#ref). *N*-Methyl-*N*-(trimethylsilyl)trifluoroacetamide (MSTFA) is a very versatile reagent because it can be used to derivatize many of the functional groups commonly found in organic compounds. It operates by attachment of a nonpolar trimethylsilyl (TMS) group (__Figure 1__). In most cases TMS derivatives have a lower boiling point than the parent compound, which allows GC analysis at a lower temperature. This reduces the possibility of thermally induced decay of compounds in the injector or on the column. For quantitative analysis the use of catalysts is recommended[2-4](https://www.sigmaaldrich.com#ref). ![Reaction of Hydroxyl Groups (Silylation)](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/protocols/analytical-chemistry/gas-chromatography/g005551.jpg "Reaction of Hydroxyl Groups (Silylation)") __Figure 1.__Reaction of Hydroxyl Groups (Silylation) with MSTFA Alcohol (R-OH) shown, but reaction also applies to R–COOH, R=NH, R–NH2, R–SH __Using Deuterated MSTFA (MSTFA-d9) for Compound Identification__ In some cases it is unclear precisely how many of the molecule’s functional groups are affected by the derivatization reaction, especially when dealing with unknown compounds. Using deuterated MSTFA (MSTFA-d9) helps to overcome this problem because the molecular mass of the MSTFA-d9 derivative increases by exactly 9 mass units for each derivatized functional group. Thus, it is possible to calculate the molecular mass of the original compound by taking the mass shift in the MSTFA/MSTFA-d9 spectra into account. __Figure 2__ shows the mass spectrum of amphetamine (α-methylphenethylamine) using GC-MS on an SLB™-5ms column. Primary and secondary amines often give an \[M-1]+l ion that can cause misinterpretation of molecular mass and failure to identify nitrogen-containing compounds. Amphetamine also generates unspecific and ambiguous major fragments; 44 m/z can match both amines and aldehydes (__Figure 2, top__). After MSTFA derivatization, the mass spectrum (__Figure 2, middle__) reveals typical fragments of TMS-derivatives (loss of methyl from TMS), while the target compound amphetamine can be calculated based on the fragment mass 192 m/z as intact molecule (192+15 m/z=207 m/z, TMS-amphetamine). Using MSTFA-d9 the 192 m/z fragment ion is shifted by 6 mass units to 198 m/z and 9 mass units to 201 m/z (__Figure 2, bottom__). The first shift indicates the attachment of one TMS group. A shift of 9 mass units is associated with the cleavage of the methyl bond in the aliphatic chain of the amphetamine structure and provides additional information about the fragmentation mechanism. ![Mass Spectra of Amphetamine (top), TMS-Amphetamine (middle), and TMS-d9-Amphetamine (bottom)](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/protocols/analytical-chemistry/gas-chromatography/g005552.gif "Mass Spectra of Amphetamine (top), TMS-Amphetamine (middle), and TMS-d9-Amphetamine (bottom)") __Figure 2.__Mass Spectra of Amphetamine (top), TMS-Amphetamine (middle), and TMS-d9-Amphetamine (bottom) Derivatization procedure:Dissolve 1 mg sample in 270 µL MSTFA (or ~10 µg sample in 25 µL MSTFA-D9). Heat for 10 minutes at 70 °C. Add 30 µL pyridine. Heat for 10 minutes at 70 °C. Dilute the solution with chloroform if necessary. column:SLB-5ms, 30 m x 0.25 mm, 0.25 µm (Product No. [28471-U](https://www.sigmaaldrich.com/US/en/product/supelco/28471u)) oven:40 °C to 300 °C in 26 min (10 min) carrier gas:helium, 1 mL/min __Figure 3__ shows the effect of MSTFA derivatization on the resultant chromatography on an SLB-5ms GC column. Despite the presence of other substances in the sample, underivatized amphetamine elutes first followed by its TMS derivative. This example shows how derivatization can be used to alter retention or selectivity to see unknown compounds that might normally coelute with the sample matrix or other components of the sample. ![GC-MS Chromatogram (TIC, EIC) of Amphetamine and TMS-Amphetamine Conditions are the same as Figure 2](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/protocols/analytical-chemistry/gas-chromatography/g005553.gif "GC-MS Chromatogram (TIC, EIC) of Amphetamine and TMS-Amphetamine Conditions are the same as Figure 2") __Figure 3.__GC-MS Chromatogram (TIC, EIC) of Amphetamine and TMS-Amphetamine Conditions are the same as Figure 2 ## Summary The majority of current GC-MS systems are equipped with single quadrupole mass analyzers with limited mass resolution. They do not offer the ability to acquire MS/MS or MSn spectra, like triple quadrupole or ion trap mass analyzers do, making them particularly amenable to the MSTFA/MSTFA-d9 derivatization described here. There is no need to change the ionization technique, such as chemical ionization, to obtain the molecular mass or run additional MS/MS experiments to elucidate the structure and composition of a fragment or molecule. Another advantage of MSTFA and MSTFA-d9 is the comparability of chromatographic properties of their corresponding derivatives. TMS and TMS-d9 derivatives elute within the same time window. Deuterated compounds may elute a few seconds earlier. Thus, researchers can use the compound retention time to find and identify a target compound even at low concentration levels. ## Materials Sorry, an unexpected error has occurred Response not successful: Received status code 500 ### References 1\. Spengler B, Lützenkirchen F, Kaufmann R. 1993. On-target deuteration for peptide sequencing by laser mass spectrometry. Org. Mass Spectrom.. 28(12):1482-1490. [https://doi.org/10.1002/oms.1210281220](https://doi.org/10.1002/oms.1210281220) 2\. Donike M. 1969. N-Methyl-N-trimethylsilyl-trifluoracetamid, ein neues Silylierungsmittel aus der reihe der silylierten amide. Journal of Chromatography A. 42103-104. [https://doi.org/10.1016/s0021-9673(01)80592-6](https://doi.org/10.1016/s0021-9673%2801%2980592-6) 3\. Donike M, Zimmermann J. 1980. Zur Darstellung von Trimethylsilyl-, Triethylsilyl- und tert.-Butyldimethylsilyl-enoläthern von Ketosteroiden für gas-chromatographische und massenspektrometrische Untersuchungen. Journal of Chromatography A. 202(3):483-486. [https://doi.org/10.1016/s0021-9673(00)91836-3](https://doi.org/10.1016/s0021-9673%2800%2991836-3) 4\. Smith R. 1994. Handbook of derivatives for chromatography?second edition K. Blau and J. Halket (editors), Wiley, Chichester. 1993. Pages: xxii + 369. £65.00. ISBN 0-471-92699-x.. Talanta. 41(10):1743-1743. [https://doi.org/10.1016/0039-9140(94)80039-1](https://doi.org/10.1016/0039-9140%2894%2980039-1) 5\. McLafferty FW, Ture?ek F. 1995. Interpretation von Massenspektren. [https://doi.org/10.1007/978-3-642-39849-0](https://doi.org/10.1007/978-3-642-39849-0) __Related Articles__ - [Allergenic Fragrance Testing – Certified Materials](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/calibration-qualification-and-validation/allergenic-fragrance-testing) - [Analysis of Alkylphenols Using New Internal Standards](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/calibration-qualification-and-validation/analysis-of-alkylphenols) - [FAME Standard for Optimizing GC System Performance](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/calibration-qualification-and-validation/fame-standard-for) - [Fast and Reliable Environmental Analysis of Aldehyde and Ketone Air Pollutants](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/calibration-qualification-and-validation/fast-and-reliable) - [New GC-MS Method for Mycotoxin Analysis](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/calibration-qualification-and-validation/new-gc-ms-method-for-mycotoxin-analysis) - [Analysis of Adulterated Lemon Essential Oil on the SLB-5ms](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/gas-chromatography/analysis-of-adulterated) - [GC Analysis of a 37-Component FAME Mix on SP™-2560](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/gas-chromatography/c-analysis-of-a-37-component-fame-mix) - [Capillary Column Ferrule Selection and Installation](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/gas-chromatography/capillary-column-ferrule) __Related Product Categories__ - [Chromatography and spectroscopy reagents](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-reagents/chromatography-spectroscopy-reagents) - [Controlled substances and illicit drug standards](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/reference-materials/pharmaceuticals-illicit-drugs-and-diagnostic-biomarkers) - [Protein degrader building blocks](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/chemical-biology/protein-degrader-building-blocks) Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars) Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions) Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices) Social Media [![LinkedIn icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/linkedin.svg)](https://www.linkedin.com/company/merck-life-science) [![X icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/x-icon.svg)](https://x.com/Merck_lifesci) [![Facebook Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/facebook.svg)](https://www.facebook.com/Merck.lifescience) [![Instagram Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/instagram.svg)](https://www.instagram.com/mercklifescience/) [![YouTube Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/youtube.svg)](https://www.youtube.com/@MerckLifeScience/featured) MilliporeSigma __Research. Development. Production.__ We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production. [![Vibrant M string](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/vibrant-m-string-1-rrvcrp.svg)](https://www.sigmaaldrich.com/US/en) [Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco) © 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies. Reproduction of any materials from the site is strictly forbidden without permission. [Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)