Attention:
We are currently upgrading the certain systems that support this website.  You may experience some functionality loss, though we expect the site to be fully functional by 9:00PM CT (US), February 24th.
We apologize for any inconvenience and appreciate your patience during this improvement process.Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Carbon Adsorbents: Physical Characteristics and Selection Guide
Carbon Adsorbents main pageThe following information is gathered to assist chemists and method developers in locating the proper specialty carbon adsorbent for their use. Additional assistance can be obtained by contacting our Technical Service group at techservice@sial.com.
Physical Characteristics of Our Specialty Adsorbents – easy-to-use chart that shows the approximate surface area, pore volume, pore diameter, and free fall density for each of our specialty carbon adsorbents
Carbon Adsorbent Cross-Reference – handy table to find replacements for many adsorbents, including some that are discontinued
Carbon Adsorbent Selection Based on Analyte Size – bed order suggestions for multi-bed air monitoring tubes
A Tool for Selecting an Adsorbent for Thermal Desorption Applications – Technical Report with detailed adsorption/desorption characteristics of fifteen of our specialty carbon adsorbents, plus nine other adsorbents
Physical Characteristics of our Specialty Carbon Adsorbents
The following information is gathered to assist chemists and method developers in locating the proper specialty carbon adsorbent for their use. Additional assistance can be obtained by contacting our Technical Service group at techservice@sial.com.
To view the approximate surface area, pore volume, pore diameter, micropore diameter, and free fall density for each of our specialty carbon adsorbents, simply click the graphic below. Also indicated is the type of adsorbent (CMS, SGPC, GCB, etc.), the shape (spherical or granular), and whether it is scalable. Included are all our Carboxen®, Carbosieve®, Graphsphere™, Carbotrap®, and Carbopack™ adsorbents. If you are unable to download this, please contact Technical Service at techservice@sial.com.
Carbon Physical Characteristics
(PDF)
Adsorbent Cross-Reference Guide
Use this easy-to-use cross-reference table to find our recommendations for non-Supelco adsorbents, including some that are discontinued.
Adsorbent Supelco Recommendation
Ambersorb 347 Carboxen 564
Ambersorb 563 Carboxen 563
Ambersorb 572 Carboxen 572
Anasorb CMS Carboxen 564, Carboxen 1000, Carbosieve S-III
Anasorb CSC Coconut charcoal
Anasorb GCB1 Carbotrap B, Carbopack B
Anasorb GCB2 Carbotrap C, Carbopack C
Anasorb 708 Chromosorb 1081
Anasorb 727 Amberlite XAD-4
Anasorb 747 Carboxen 564, Carboxen 1000, Carbosieve S-III
CarboBlack B Carbotrap B, Carbopack B
CarboBlack C Carbotrap C, Carbopack C
Carbograph 1 Carbotrap B, Carbopack B
Carbograph 2 Carbotrap C, Carbopack C
Carbograph 3 Carbotrap F, Carbopack F
Carbograph 4 Carbopack Z
Carbograph 5 Carbotrap X, Carbopack X
Carbosphere Carboxen 1000
Graphpac-GC Carbotrap B, Carbopack B
Purosieve Carboxen 1000
Spherocarb Carboxen 1000
Tenax GC Tenax TA1
1 Supelco offers this adsorbent, but is not the manufacturer.
Carbon Adsorbent Selection Based on Analyte Size
To assist with the selection of our specialty carbon adsorbents, this table below lists some general guidelines based on analyte size. For multi-bed tubes, use the weaker adsorbent in front of the stronger adsorbent to protect it (this will keep analytes from adsorbing on a bed too strong, resulting in poor analyte release during desorption). For example, use Carbotrap C in front of Carbotrap B.
Relative Analyte Size1 Adsorptive
Strength
Recommended Adsorbents (listed highest to lowest surface area)
C20+ Weakest
|
Strongest
Carbotrap F, Carbopack F
C12-C20 Carbotrap C, Carbopack C
C9-C14 Carbotrap Y, Carbopack Y
C5-C12 Graphsphere 2027, Graphsphere 2029, Carbotrap B, Carbopack B, Graphsphere 2016, Graphsphere 2017
C3-C9 Carbotrap X, Carbopack X, Carbopack Z
C2-C5 Carboxen 1012, Carboxen 1034, Carboxen 1000, Carboxen 1008, Carboxen 1026, Carbosieve G, Carboxen 1005, Carboxen 572, Carbosieve S-II, Carboxen 1003, Carbosieve S-III, Carboxen 1032, Carboxen 1030, Carboxen 1006, Carboxen 1018, Carboxen 1010, Carboxen 1021, Carboxen 563, Carboxen 1001, Carboxen 569, Carboxen 1033, Carboxen 564
1 Analyte size relative to n-alkanes. Consider all atoms, not just carbon. For example, even though 1,2-dichloroethane is a C2, the two chlorine atoms give it a relative size between C4 and C5.
A Tool for Selecting an Adsorbent for Thermal Desorption Applications
(PDF)
Detailed adsorption/desorption characteristics of fifteen of our specialty carbons plus nine other adsorbents, depicted in twenty-four easy-to-follow color-coded tables, are shown in A Tool for Selecting an Adsorbent for Thermal Desorption Applications (T402025 HTA). To download your no-charge copy of this Technical Report, simply click the graphic of the report cover. If you are unable to download this, contact Technical Service at techservice@sial.com.