Chemizorb® Absorbents
Absorbents for Spilled Liquids
Quick and safe cleaning with Chemizorb®
When spills happen, Chemizorb® absorbents are the quick, safe and environmentally friendly way to remove hazardous, aggressive or unpleasant liquids. Our fast-acting absorbents are made of porous mineral or synthetic copolymers that are chemically inert, and capable of absorbing up to 400% of their own weight in liquid. We offer a choice of absorbents for a variety of spills.
“All-round” Chemizorb® powder or granules
  • Suitable for aqueous solutions (acids, alkalis), organic solvents and viscous oils
  • Excellent absorbance capacity
  • Powder offers higher absorbance
  • Granules are easier to dose and more suitable for draughty rooms or outdoors
“Specialist” Chemizorb® absorbents for alkalis, acids and hydrofluoric acid
  • Contain water-soluble neutralizers 
  • pH indicators keep track of neutralization process
  • Aggressive acids and alkalis removed more safely and efficiently
“All-in-one” Chemizorb® Hg set for mercury
  • Set contains all reagents and auxiliaries needed
  • Safe, complete removal of mercury drops or traces 
  • Sufficient for decontamination of approx. one square meter 
Materials
If you have trouble finding the products you're looking for, please contact our customer support for assistance.