Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Acids in Safebreak Bottles
Just in Case
Glass bottles are ideal for storing acids because they are inert and offer a long shelf life. The only disadvantage of glass is that it can break and cause serious injury, acid spills, contamination or consequential damage. Fortunately, you can avoid all this by using acids in Safebreak bottles.
Safebreak is a robust glass bottle coated with polyethylene (PE). Should the bottle fall and break, all acid and glass splinters are safely retained within the polyethylene coating, thus protecting you from any risk.
Now the Safebreak bottles are available in 3 different sizes: 0.5 liter, 1 liter and 2.5 liter.
Hydrochloric acid, sulfuric acid, and ortho-phosphoric acid in Safebreak bottles
Benefits
  • Safe: Should breakage occur, all acid and glass splinters are retained by the PE coating
  • Ecological: Thanks to the PE coating, Safebreak bottles can be recycled without affecting the environment
  • Durable: Same long shelf life as conventional glass bottles
  • Tightly closed: Safebreak bottles are fitted with our exclusive S40 screw cap made of PP with an integrated PTFE component
  • Economical: Just as recyclable as conventional glass bottles
Ordering Information
Enjoy added safety with the following acids in Safebreak bottles:  
Product Pack size Packaging material Ord. No.
Acetic acid (glacial) 100% anhydrous for analysis EMSURE® ACS,ISO,Reag. Ph Eur 0.5 L Safebreak bottle 1000630510
1 L Safebreak bottle 1000631010
2,5 L Safebreak bottle 1000632510
Hydrochloric acid fuming 37% for analysis EMSURE® ACS,ISO,Reag. Ph Eur 0.5 L Safebreak bottle 1003170510
1 L Safebreak bottle 1003171010
2,5 L Safebreak bottle 1003172510
Nitric acid 65% for analysis EMSURE® ISO 0.5 L Safebreak bottle 1004560510
1 L Safebreak bottle 1004561010
2,5 L Safebreak bottle 1004562510
Nitric acid 69% for analysis EMSURE® ACS,Reag. Ph Eur 0.5 L Safebreak bottle 1017990510
1 L Safebreak bottle 1017991010
2,5 L Safebreak bottle 1017992510
ortho-Phosphoric acid 85% for analysis EMSURE® ACS,ISO,Reag. Ph Eur 0.5 L Safebreak bottle 1005730510
2,5 L Safebreak bottle 1005732510
Perchloric acid 70-72% for analysis EMSURE® ACS,ISO,Reag. Ph Eur 0.5 L Safebreak bottle 1005190510
1 L Safebreak bottle 1005191010
2,5 L Safebreak bottle 1005192510
Sulfuric acid 95-97% for analysis (max. 0.005 ppm Hg) EMSURE® ACS,ISO,Reag. Ph Eur 0.5 L Safebreak bottle 1007320510
2,5 L Safebreak bottle 1007322510
Sulfuric acid 95-97% for analysis EMSURE® ISO 0.5 L Safebreak bottle 1007310510
1 L Safebreak bottle 1007311010
2,5 L Safebreak bottle 1007312510
Sulfuric acid 98% for analysis EMSURE® 0.5 L Safebreak bottle 1120800510
2,5 L Safebreak bottle 1120802510
Related Links
Learn more about acids in Safebreak bottles: