We offer a comprehensive portfolio of inorganic salts in two quality grades: EMSURE® and EMPLURA®. Our salts are manufactured under strictly controlled conditions mostly in our facilities at Darmstadt, Germany. Their outstanding analytical purity, especially for the EMSURE® grade, makes them ideal for both qualitative and quantitative inorganic analysis. They meet or exceed international standards such as ISO, ACS and Reag. Ph Eur and come with an extensive Certificate of Analysis.
Our premium salts designated "for analysis EMSURE® ACS, Reag. Ph Eur" are specified for pharmacopoeia analysis and fully comply to the requirements of both the European and U.S. pharmacopoeias. Thus, quality assurance laboratories in the pharmaceutical industry benefit from both the international acceptance of these reagents and the security of analytical laboratory auditing.
Due to our broad knowledge about producing salts, we have improved the flowability of various products to make their use more convenient.
Use our EMPLURA® grade salts for preparative lab work and basic applications and profit from adequate specifications with the most common parameters and convenient pack sizes.
If you have trouble finding the products you're looking for, please contact our customer support for assistance.