Solvents for Classical Analysis, Production and Purification
Just the Right Solvent for Your Application
For each of your applications, you want to use solvents that perfectly fit your needs. That’s why we strive to tailor our comprehensive range of solvents to customer requirements. With solvents from us, you are on the safe side.
EMPARTA® is a high quality solvent that fulfills the requirements of the ACS and can be used for a wide range of analytical applications. If you want even more, we offer you EMSURE®. With more test parameters and improved values for purity, water content and evaporation residue, EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications including regulated and highly demanding applications. Furthermore, EMSURE® is not only compliant to ACS, but also to Reag. Ph Eur and ISO regulations.
EMPLURA® solvents are a cost-effective alternative for preparative lab work, production and cleaning. For many applications, e.g. DNA/RNA synthesis, peptide and organic synthesis, dried solvents are essential. SeccoSolv® ready-to-use dried solvents offer highest purity, high and constant dryness – and more flexibility thanks to a wide range of packaging solutions including our innovative SeccoSept® septum seal cap for best quality protection.
Our Prepsolv® solvents are tailored to the requirements of preparative chromatography to facilitate scale-up from analytical to preparative separations. With their extremely low evaporation residue (< 1 mg/l) and low water content, they ensure optimal protection for columns.
If you have trouble finding the products you're looking for, please contact our customer support for assistance.