Gas Chromatography Solvents
Our GC solvent qualities are ideal for all gas chromatography applications. These solvents are specially developed and tested for ECD, FID and MS detection to ensure a maximum signal-to-noise ratio in the largest specified retention time range. Our solvents for headspace gas chromatography are developed specifically for the analysis of residual solvents in drug substances, excipients, and drug products according to Ph. Eur. and USP. High purity is provided by specially designed production processes for accurate, reliable and reproducible analytical results.
SupraSolv® GC Solvents Specifications
SupraSolv® solvents SupraSolv® Headspace
for analysis of residual solvents
SupraSolv® hypergrade for organic trace analysis SupraSolv®
for GC-ECD/FID
SupraSolv®
for GC-MS
Suitability Tested GC Headspace technique Electron capture detector, flame ionization detector and mass spectroscopy Electron capture detector and flame ionization detector GC-MS/MS
Descriptions Free from contaminants could possibly interfere with GC analysis by coeluting with peaks of interest in the sample Narrow specification, the highest purity, and larger retention time range; low boiling substances can be reliably detected; the permissible concentration of interference signals within the retention time range is lower Meet EPA Method 508 and the stringent demands of capillary gas chromatography; glass-distilled solvents resulting in exceptional quality and low residue levels Undergo rigorous purification procedures and assure high chemical purity; lowest impurity profile and minimal signal-to-noise ratio in specification
Specification Every residual solvent of class 1 acc. CH ≤ 1 µg/g
Every residual solvent of class 2 acc. CH ≤ 10 µg/g
Every residual solvent of class 3 acc. CH ≤ 50 µg/g
≤ 1 ng/mL compared to Tetrachloromethane GC-ECD
≤ 2 pg/mL compared to Lindane GC-ECD
≤ 2 ng/mL compared to n-Tetradecane GC-FID
≤ 2 ng/mL compared to n-Tetradecane GC-MS
3 pg/mL compared to Lindane GC-ECD
≤ 3 ng/mL compared to n-Tetradecane GC-FID
≤ 3 ng/mL compared to n-Tetradecane GC-MS
Benefits Extremely high purity resulting in a very low baseline
Microfiltered (0.2 μm)
Packaged under inert gas for longer shelf life
Meet USP, Ph. Eur. and ICH guidelines
Extremely low residual impurities
Multi-purpose grade
Very high purity
Improves column lifetime due to extremely low content of non-volatile matter
low residue levels
Very high purity
Improves column lifetime due to extremely low content of non-volatile matter
Applications Residual solvents analysis in drug substances and products acc. Ph Eur & USP Pesticide analysis
Organic residue analysis
Volatile halogenated hydrocarbons in water
Polychlorinated biphenyls (PCB) in water
Detection of Acrylamide in food
BTX analysis (Benzene, Toluene, Xylene) detection
Hydrocarbon-oil-index in water
Determination of emissions in car cockpit materials
Analysis of Dioxins and Furans (PCDD/PCDF)
Analysis of polycyclic aromatic hydrocarbons
Pesticide analysis acc. QuEChERS
Determination of phthalates in plastics
SupraSolv®solvents for gas chromatography ECD and FID
Solvent Available Pack Sizes Product Number
Acetone 1 L GL ,2.5 L GL ,4 L GL ,30 L ST 100012
Acetonitrile 1L GL ,2.5L GL 100017
tert-Butyl methyl ether 1 L GL ,2.5 L GL 101995,
Chloroform, stabilized 1 L GL ,2.5 L GL 102432
Cyclohexane 1 L GL ,2.5 L GL ,4 L GL ,10 L ST 102817
Dichloromethane, stabilized 1 L GL ,2.5 L GL ,4 L GL ,10 L ST 106054
Diethyl ether, stabilized 1 L GL ,2.5 L GL ,4 L GL 100931
N,N-Dimethylformamide 1 L GL ,2.5 L GL 110983
Ethanol 1 L GL ,2.5 L GL 102371
Ethyl acetate
1 L GL ,2.5 L GL ,4 L GL ,10 L ST,30 L ST
110972
n-Hexane 1 L GL ,2.5 L GL ,4 L GL ,10 L ST,30 L ST 104371
Isohexane 2.5 L GL 104340
Isooctane 1 L GL ,2.5 L GL 115440
Methanol 1 L GL ,2.5 L GL,4 L GL 106011
n-Pentane 1 L GL ,2.5 L GL 100882
Petroleum benzine (40 - 60 °C) 1 L GL ,2.5 L GL ,4 L GL ,10 L ST,30 L ST 101772
2-Propanol 1 L GL ,2.5 L GL 100998
Toluene 1 L GL ,2.5 L GL ,4 L GL 108389
Water 1 L GL ,2.5 L GL 102699
GL = glass bottle I ST = stainless steel barrel I * = sum of hexane isomers + methyl cyclopentane (GC) ≥ 99.8 %
GC-ECD (retention range 1,2,4-Trichlorobenzene to Decachlorobiphenyle individual signals (Lindane standard)) :, 3 pg/mL
GC-FID (retention range n-Undecane to n-Tetracontane individual signals (n-Tetradecane standard)) :, 3 ng/mL
SupraSolv® solvents for gas chromatography MS
Solvent Available Pack Sizes Product Number
Acetone 1L GL, 2.5L GL 100658
Acetonitrile 1L GL, 2.5L GL 100665
Cyclohexane 1L GL, 2.5L GL 100667
Dichloromethane, stabilized 1L GL, 2.5L GL 100668
Ethyl acetate 1L GL, 2.5L GL 100789
n-Hexane 1L GL, 2.5L GL 100795
Methanol 1L GL, 2.5L GL 100837
Toluene 1L GL, 2.5L GL 100849
Water 1L GL, 2.5L GL 103702
Water, ACS reagent, for ultratrace analysis 1L GL 14211
1-Methyl-2-pyrrolidinone 1L GL 43729
Methyl pivalate 1L GL 52596
GL = glass bottle I * = sum of hexane isomers + methyl cyclopentane (GC) ≥ 99.8 %
GC-MS (retention range n-Undecane to n-Tetracontane; scanning area 30 - 600 amu individual signals (n-Tetradecane standard)) ≤ 3 ng/mL
SupraSolv® headspace for the analysis of residual solvents according to ICH, Ph Eur and USP
Solvent Available Pack Sizes Product Number
1-Methyl-2-pyrrolidone 0.5L GL, 1L GL, 2.5L GL 102497
Benzylalcohol 1L GL, 2.5L GL 102695
N,N-Dimethylacetamide 0.5L GL, 1L GL 100399
N,N-Dimethylformamide 1L GL, 2.5L GL 100202
Dimethyl sulfoxide 1L GL, 2.5L GL 101900
Water 1L GL, 2.5L GL 100577
GL = glass bottle
Every residual solvent of class 1 acc. CH ≤ 1 μg/g
Every residual solvent of class 2 acc. CH ≤ 10 μg/g
Every residual solvent of class 3 acc. CH ≤ 50 μg/g
SupraSolv® hypergrade solvents for organic trace analysis
Solvent Available Pack Sizes Product Number
Dichloromethane 1L GL 106454
n-Hexane 1L GL, 2.5L GL 104369
n-Pentane 1L GL, 2.5L GL 107288
Petroleum benzine (40 - 60°C) 1L GL, 2.5L GL 116740
GL = glass bottle
* Sum of hexane isomers + methylcyclopentane (GC) > 99.9 %
GC-ECD (retention range Dichloromethane to 1,2,4-Trichlorobenzene individual signals (Tetrachloromethane standard)) ≤ 1 ng/mL
GC-ECD (retention range 1,2,4-Trichlorobenzene to Decachlorobiphenyle individual signals (Lindane standard)) ≤ 2 pg/mL
GC-FID (retention range n-Undecane to n-Tetracontane individual signals (n-Tetradecane standard)) ≤ 2 ng/mL
GC-MS (retention range n-Undecane to n-Tetracontane; scanning area 30 - 600 amu individual signals (n-Tetradecane standard) ≤ 2 ng/mL