Anhydrous Solvents for Organic and Organometallic Chemistry
Organic and organometallic chemistry
Many well-known reactions require water-free conditions to initiate or complete the reaction. Failure to include anhydrous solvents can result in low product yield or reaction failure. As the market leader of anhydrous solvents, MilliporeSigma offers high quality solvents with extremely low water levels, specifically manufactured for all facets of moisture-sensitive organic and organometallic chemistry.
Grignard reaction
Grignard reactions require extremely low water and air conditions due to the strong nucleophilicity of the Grignard reagent. Due to this condition requirement, it is important to perform the reaction with high quality ether-based anhydrous solvents (or aprotic solvents).
Grignard reaction
Cat no. Product description Greener alternative
227056 N,N-Dimethylformamide, anhydrous, 99.8%  
328634 1-Methyl-2-pyrrolidinone anhydrous  
401757 Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free  
244511 Toluene, anhydrous, 99.8%  
675970 Cyclopentyl methyl ether, contains 50 ppm BHT as inhibitor, anhydrous, ≥99.9% Greener Alternatives
414247 2-Methyltetrahydrofuran, anhydrous, ≥99.0%, contains 250 ppm BHT as stabilizer Greener Alternatives
Vilsmeier–Haack reaction
One of the more common reactions for aryl aldehyde formation (or formylation) is the Vilsmeier-Haack reaction. This reaction begins with the formation of Vilsmeier reagent, which requires high-quality anhydrous solvents.
Vilsmeier–Haack reaction
Cat no. Product description
270997 Dichloromethane anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
227056 N,N-Dimethylformamide, anhydrous, 99.8%
240664 1,2-Dichlorobenzene anhydrous, 99%
Friedel–Crafts reaction
A typical electrophilic aromatic substitution reaction that involves both alkylation and acylation; it is one of the more important reactions for organic and biological synthesis. The use of anhydrous solvents is highly recommended to prevent any unnecessary side reaction.
Friedel–Crafts reaction
Cat No. Product Description
284505 1,2-Dichloroethane, anhydrous, 99.8%
271004 Acetonitrile, anhydrous, 99.8%
296996 Methyl acetate, anhydrous, 99.5%