Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Solvents for Gas Chromatography SupraSolv®
Dedicated Solvents for Your Gas Chromatography Applications
SupraSolv® solvents are developed specially for sample preparation in gas chromatography. Ideally suited for all GC applications including sensitive detection tasks in residue and environmental analysis, SupraSolv® solvents offer you the security and reliability needed for today’s gas chromatography applications.
No matter if you use ECD, FID or MS – our comprehensive portfolio of GC solvents offers a dedicated product quality for your specific application and detection method. Our SupraSolv® ECD and FID quality is specially developed and tested for ECD (Electron Capture Detector) and FID (Flame Ionization Detector). Typical applications include the determination of polychlorinated biphenyls (PCB) in water and soil or pesticides in fruits and vegetables. SupraSolv® MS is dedicated for use in gas chromatography coupled with mass spectrometric detection, e. g. the analysis of dioxins and furans (PCDD/PCDF) in food and water samples or the determination of PAH (polycyclic aromatic hydrocarbons) in food.
Both SupraSolv® qualities are carefully tested with these specific detectors, showing highest purity and industry-leading performance. Their high quality and batch consistency allows reliable, accurate and reproducible results without the need for costly purification and repeated analysis. As a result, they make your work easier, more efficient and more economical.
Materials
If you have trouble finding the products you're looking for, please contact our customer support for assistance.