Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Solvents for Headspace Gas Chromatography SupraSolv®
Solvents for Headspace GC Analysis of Residual Solvents According to Ph Eur and USP
SupraSolv® headspace is the range of solvents specially designed for headspace gas chromatography. Headspace GC is the preferred method for the analysis of residual solvents in actives, excipients, and drug products according to Ph Eur and USP.
Accurate analysis with headspace gas chromatography requires the use of very pure solvents with extremely low concentrations of the defined residual solvents. We ensure this high purity through special, state-of-the-art production processes, and we specify for our high-purity SupraSolv® headspace solvents the concentrations of all the residual solvents classified in the relevant ICH guideline Q3C "Impurities: Guideline for Residual Solvents".
Moreover, we perform application tests on every single batch of SupraSolv® headspace – thus we are able to consistently deliver you the reliability, accuracy and analytical safety you need.
Materials
If you have trouble finding the products you're looking for, please contact our customer support for assistance.