Digital Test Strip Readout with MQuant® StripScan
Analyze your MQuant® test strips with your smartphone for fast, convenient, and precise results
Test the simplest way to determine pH or concentrations of chemicals in aqueous solutions. This low-priced and easy-to-use analytical detection system provides reliable quantification of water, food & beverage samples for customers from research, industry, environment and diagnostics.
  • Quick, easy, and reliable
  • Superior precision – finer gradation of results
  • Convenient data analysis
Read out your MQuant® test strips in seconds with the StripScan app, and analyze, graph and share your data measurements with the StripScan Web application!
Chemical residues are especially important to monitor after disinfection control as their presence may cause adverse health effects or affect the quality of the final product. Join our webinar to find out how to quickly analyze chemical disinfectant parameters to ensure the safety of your production line after disinfection & the different methods available
Do you want to contribute to a greener future one strip at a time? Let us know how we can do more to reduce our carbon footprint while still offering high-quality test strips! Take the survey!
Three components for your digital readout test system
MQuant® StripScan –
the FREE mobile App
The tool for digital test strip readout
• Make your measurements
  with MQuant® StripScan
  and effortlessly manage,
  visualize and share your results.
• StripScan allows you to
  digitally read out your test
  results everywhere.
The MQuant® StripScan App is available for free in the App Store,
Available all current iOS versions from iOS 10 and newer
MQuant® StripScan Reference Cards
The color reference for precise results
• The MQuant® StripScan Reference
  Card ensures reliable and precise
  results by providing an external
  color standard.
• The brilliant and colorfast
  reference colors allow for robust
  measurement results even under
  changing lighting conditions.
MQuant® StripScan Reference Card for pH (103736):
Analyze your pH 0-14 test strips (109535) with this reference card and with the free MQuant® StripScan app.
MQuant® StripScan Reference Card for Nitrate (103733):
This reference card allows for the digital readout of nitrate test strips 110020 and 110092, at a concentration range between 0 mg/L and 500 mg/L nitrate. Use with the free MQuant® StripScan app.
MQuant® Test Strips
Your quick, easy and low-priced chemical analysis tools
MQuant® pH Indicator Test Strips (109535):
Due to our unique manufacturing process, pH indicator strips do not bleed, so that that measurements can be made directly in the sample without contaminating it. The pH tests are suited for environmental analyses and in industrial in-process controls.
MQuant® Nitrate Test Strips (110020):
The MQuant® nitrate test can be used for monitoring of drinking
water, process water and wastewater, as well as in aquarium water where thresholds can quickly be exceeded. It can also be used for determining nitrate in food, e.g. potatoes.
MQuant® StripScan Web
Data analysis platform
Sign in to StripScan web for advanced data analysis options. Group your data into experiments and graph, share, delete and export your data. Convenient and intuitive, StripScan Web allows you to take your data analysis further. Explore the free platform here
How to use StripScan
Start the app and get your MQuant® test strip and Reference card ready
• Step 1: Test your sample with the
  test strip
• Step 2: Choose the corresponding
  parameter in the app and await
  countdown
• Step 3: Place your test strip on
  the Reference Card and take an
  image with the automatic app
  acquisition mechanism
• Step 4: Read out the result and
  compare it with your data
Application Notes
• Fields of application for mobile
  analyses: water or wastewater
  testing, disinfection control, food &
  beverages, quality control and in
  process monitoring
With over 300 application notes
  for photometry and
  reflectometry
  (including MQuant® StripScan),
  find the ideal method to support
  your production process and
  analyses.