Supelco® SmartChemicals for Titration
Titration goes digital—Connect your SmartChemicals to your Titrator
Connect your SmartChemicals with your Titrator
A new titration technology, using Supelco® SmartTitrants & Supelco® SmartStandards transfers data seamlessly to the titrator. Just a brief touch on the SmartChemical RFID (Radio Frequency Identification) tag saves time, reduces errors and always ensures maximum data integrity.
Seamless transfer of titration data via RFID tag
Using SmartChemicals eliminates time-consuming steps and errors by transferring data wirelessly and instantly. Titripur® volumetric solutions, Certipur® volumetric standards and all Aquastar® Karl Fischer titrants and standards are embedded with an RFID tag on the label. All relevant data from the Certificate of Analysis are stored on the RFID tag of the SmartChemical.
Seamless Data Transfer with Supelco® SmartChemicals
Just a simple touch transfers all data in seconds
All information needed for method execution, result calculation and documentation is seamlessly transferred
  • Product name
  • Article number
  • Lot/batch no
  • Concentrations
  • Date of release
  • Shelf life
When using standards, the following additional information is also transferred:
  • Molecular weight
  • Supplier name
  • Compliance according to pharmacopeias and ISO
  • Measurement uncertainty
Supelco® SmartTitrants & Supelco® SmartStandards
Initial opening date – stored on the tag
Previously opened chemicals are also recognized, the initial opening date is stored in the RFID tag and is shown on the touchscreen. Expired chemicals are automatically locked from use.
Four benefits using SmartChemicals for your titration
  • Secure data transfer—ensures complete and correct reagent & standard data transfer in the titration software
  • Ease of use—intuitive operation and convenient data transfer in one touch
  • Extended quality management—titer determination, shelf life, compliance data, initial opening date, release date
  • Improved efficiency—fast data transfer - safes time - no manual writing and no four-eyes principle necessary
SmartChemicals are compatible with METTLER TOLEDO Titrators
Compact Titrator
  • G20S
  • G10S
  • V30S
  • V20S
  • V10S
Mettler Toledo Compact Titrator
Excellence Titrators
  • T9
  • T7
  • T5
Mettler Toledo Excellence Titrator
External SmartChemical Reader
SmartChemical titration data reader
Video: Titration goes Digital
Supelco® SmartChemicals - Product List
All products ending with a “3” as last digit, are embedded with an RFID tag
   To order contact us
Article No. Name  Packsize/Qty Packaging
Titripur® Volumetric Solutions certified according to ISO 17025    
1022771003 Titripur® Ammonium cer(IV)nitrate solution 0.1 mol (0.1 N) 1 L PE bottle
1090791003 Titripur® Ammonium thiocyante solution 0.1 mol (0.1N) 1 L PE bottle
1603161003 Titripur® Bromide Bromate  solution  0.05 mol (0.1N) 1 L PE bottle
1090921003 Titripur® Cer (IV) sulfate solution 0.1 mol (0.1 N) 1 L PE bottle
1027841003 Titripur® Cooper sulfate solution 0.1 mol (0.1 N) 1 L PE bottle
1091641003 Titripur® Hanus solution 0.1 mol for determination of iodine number 1 L Glass bottle
1154801003 Titripur® Hyamine1622 solution for determination of anionic tensides 0.004 mol 1 L  PE bottle
1003261003 Titripur® Hydrochloric acid solution in 2-propanol 0.1 mol (0.1 N) 1 L Glass bottle
1602381003 Titripur® Hydrochloric acid solution 0.01 mol (0.01 N) 1 L PE bottle
1090601003 Titripur® Hydrochloric acid solution 0.1 mol (0.1 N) 1 L PE bottle
1090609013 Titripur® Hydrochloric acid solution 0.1 mol (0.1 N) 10 L Titripac
1090604003 Titripur® Hydrochloric acid solution 0.1 mol (0.1 N) 4 L Titripac
1131369013 Titripur® Hydrochloric acid solution 0.3571 mol (1/2.8 N) 10 L Titripac
1090581003 Titripur® Hydrochloric acid solution 0.5 mol (0.5 N) 1  L PE bottle
1090584003 Titripur® Hydrochloric acid solution 0.5 mol (0.5 N) 4 L  Titripac
1090571003 Titripur® Hydrochloric acid solution 1.0 mol (1.0 N) 1 L PE bottle
1090574003 Titripur® Hydrochloric acid solution 1.0 mol (1.0 N) 4 L Titripac
1090579013 Titripur® Hydrochloric acid solution 1.0 mol (1.0 N) 10 L Titripac
1090631003 Titripur® Hydrochloric acid solution 2.0 mol (2.0 N) 1 L PE bottle
1131349013 Titripur® Hydrochloric acid solution 3.571 mol (1/0.28N) 10 L  Titripac
1603281003 Titripur® Hydrochloric acid solution 4.0 mol (4.0 N) 1 L PE bottle
1090991003 Titripur® Iodine solution 0.05 mol (0.1 N) (not for North America) 1 L Glass bottle
1090991073 Titripur® Iodine solution 0.05 mol (0.1 N) (for North America only) 1 L Glass bottle
1091431003 Titripur® Mercury(II)nitrate solution 0.05 mol (0.1 N) 1 L PE bottle
1603291003 Titripur® Oxalic acid solution 0.005 mol (0.01 mol) 1 L PE bottle
1090651003 Titripur® Perchloric acid solution in water free acetic acid 0.1 mol  (0.1 N) 1 L Glass bottle
1603081003 Titripur® Potassium bromate solution 1/60 mol  (0.1 N) 1 L PE bottle
1603311003 Titripur® Potassium chloride solution 0.1 mol  (0.1 N) 1 L PE bottle
1091191003 Titripur® Potassium dichromate solution 0.02 mol   1 L Glass bottle
1091181003 Titripur® Potassium dichromate solution 1/24 mol (0.25 N) 1 L  Glass bottle
1091121003 Titripur® Potassium hydroxide solution 0.1 mol (0.1 N) 1 L PE bottle
1091151003 Titripur® Potassium hydroxide solution in ethanol 0.1 mol (0.1 N) 1 L Glass bottle
1055441003 Titripur® Potassium hydroxide solution in 2-propanol 0.1 mol (0.1 N) 1 L PE bottle
1091141003 Titripur® Potassium hydroxide solution in ethanol 0.5 mol (0.5 N) 1 L Glass bottle
1093511003 Titripur® Potassium hydroxide solution in methanol 0.5 mol (0.5 N) 1 L Glass bottle
1603341003 Titripur® Potassium hydroxide solution in methanol 1 mol (1 N) 1 L Glass bottle
1091081003 Titripur® Potassium hydroxide solution 1 mol (1 N) 1 L PE bottle
1091071007 Titripur® Potassium hydroxide solution max 0.4 ppm Ca 1 mol (1 N) 1 L PE bottle
1091211003 Titripur® Potassium permanganate solution standarized with sodium thiosulfate 0.02 mol (0.1 N) 1 L Glass bottle
1091221003 Titripur® Potassium permanganate solution standarized with oxalate 0.02 mol (0.1 N)  1 L Glass bottle
4801602503 Titripur® Potassium permanganate solution  0.05 mol (0.25 N) 2.5 L Glass bottle
1117181003 Titripur® Silver nitrate solution 0.05 mol (0.05 N) 1 L PE bottle
1090811003 Titripur® Silver nitrate solution 0.1 mol (0.1 N) 1 L PE bottle
1090814003 Titripur® Silver nitrate solution 0.1 mol (0.1 N) 4 L Titripac
1090819013 Titripur® Silver nitrate solution 0.1 mol (0.1 N) 10 L Titripac
1062771003 Titripur® Sodum arsenit solution 0.05 mol (0.1 N) 1 L PE bottle
1603101003 Titripur® Sodum carbonate solution 0.05 mol (0.1 N) 1 L PE bottle
1603091003 Titripur® Sodium hydroxide solution  0.01 mol (0.01 N) 1 L PE bottle
1603094003 Titripur® Sodium hydroxide solution  0.01 mol (0.01 N) 4 L Titripac
1091420503 Titripur® Sodium hydroxide solution  0.02 mol (0.02 N) 0.5 L PE bottle
1091411003 Titripur® Sodium hydroxide solution  0.1 mol (0.1 N) 1 L PE bottle
1091414003 Titripur® Sodium hydroxide solution  0.1 mol (0.1 N) 10 L Titripac
1091419013 Titripur® Sodium hydroxide solution  0.1 mol (0.1 N) 4 L Titripac
1091401003 Titripur® Sodium hydroxide solution  0.2 mol (0.2 N) 1 L PE bottle
1091404003 Titripur® Sodium hydroxide solution  0.2 mol (0.2 N) 10 L Titripac
1091391003 Titripur® Sodium hydroxide solution 0.25 mol (0.25 N) 1 L PE bottle
1091399013 Titripur® Sodium hydroxide solution 0.25 mol (0.25 N) 10 L Titripac
1091381003 Titripur® Sodium hydroxide solution 0.5 mol (0.5 N) 1 L PE bottle
1091384003 Titripur® Sodium hydroxide solution 0.5 mol (0.5 N) 4 L Titripac
1091389013 Titripur® Sodium hydroxide solution 0.5 mol (0.5 N) 10  L Titripac
1091371003 Titripur® Sodium hydroxide solution 1 mol (1 N) 1 L PE bottle
1091374003 Titripur® Sodium hydroxide solution 1 mol (1 N) 4 L Titripac
1091379013 Titripur® Sodium hydroxide solution 1 mol (1 N) 10  L Titripac
1091361003 Titripur® Sodium hydroxide solution 2 mol (2 N) 1 L PE bottle
1603181003 Titripur® Sodium thiosulfate solution  0.01 mol (0.01 N) 1 L PE bottle
1091471003 Titripur® Sodium thiosulfate solution  0.1 mol (0.1 N) 1 L PE bottle
1091474003 Titripur® Sodium thiosulfate solution  0.1 mol (0.1 N) 4 L Titripac
1091479013 Titripur® Sodium thiosulfate solution  0.1 mol (0.1 N) 10 L Titripac
1090741003 Titripur® Sulfuric acid solution 0.05 mol (0.1 N) 1 L  PE bottle
1090749013 Titripur® Sulfuric acid solution 0.05 mol (0.1 N) 10 L Titripac
1090731003 Titripur® Sulfuric acid solution 0.25 mol (0.5 N) 1 L PE bottle
1090721003 Titripur® Sulfuric acid solution 0.5 mol (1 N) 1 L PE bottle
1090724003 Titripur® Sulfuric acid solution 0.5 mol (1 N) 4 L Titripac
1090729013 Titripur® Sulfuric acid solution 0.5 mol (1 N) 10 L Titripac
4803641003 Titripur® Sulfuric acid solution 2.5 mol (5 N) 1 L PE bottle
1091621003 Titripur® Tetra-n-butyl-ammonium hydroxide solution in 2-propanol/methanol 0.1 mol (0.1 N) 1 L Glass bottle
1084311003 Titripur® Titriplex III Solution 0.1 mol  (0.05 N) (not for North America) 1 L PE bottle
1084311073 Titripur® Titriplex III Solution 0.1 mol  (0.05 N) (for North America only) 1 L PE bottle
1084314003 Titripur® Titriplex III Solution 0.1 mol  (0.05 N) (not for North America) 4 L Titripac
1084319013 Titripur® Titriplex III Solution 0.1 mol  (0.05 N) (not for North America) 10 L Titripac
1084201003 Titripur® Titirplex solution B 10 mgCaO/L = 1 ml 1 L PE bottle
1084209010 Titripur® Titirplex solution B 10 mgCaO/L = 1 ml 10 L Titripac
1084501003 Titripur® Trifluoromethansulfonic acid in anhydrous acetic acid 0.1 mol 1 L Glass bottle
1091631003 Titripur® Wijs solution (iodine monochloride in acetic acid) 0.1 mol  1 L Glass bottle
1088791003 Titripur® Zinc sulfate solution 0.1 mol (0.1 N) 1 L PE bottle
Certipur® Certified Volumetric Standards according to ISO 17034
1024000083 Certipur® Potassium hydrogen phthalate CRM 80 g Glass bottle
1024010063 Certipur® Benzoic acid CRM 60 g Glass bottle
1024020083 Certipur® Iron(II) ethylenediammonium sulfate CRM 80 g Glass bottle
1024030083 Certipur® Potassium dichromate CRM 80 g Glass bottle
1024040103 Certipur® Potassium iodate CRM 100 g Glass bottle
1024050083 Certipur® Sodium carbonate CRM 80 g Glass bottle
1024060083 Certipur® Sodium chloride CRM 80 g Glass bottle
1024070063 Certipur® Di-Sodium oxalate CRM 60 g Glass bottle
1024080083 Certipur® TRIS(Hydroxymethyl)aminomethane CRM 80 g Glass bottle
1024090103 Certipur® Zinc CRM 100 g Glass bottle
1024100053 Certipur® Calcium carbonate CRM 50 g Glass bottle
Aquastar® Karl Fischer Titrants according to ISO 17034
1880011003 Aquastar® CombiTitrant 1  one component reagents 1 mg H2O/ml (not for North America) 1 L Glass bottle
1880011043 Aquastar® CombiTitrant 1  one component reagents 1 mg H2O/ml (for North America only) 1 L Glass bottle
1880021003 Aquastar® CombiTitrant 2  one component reagents 2 mg H2O/ml (not for North America) 1 L Glass bottle
1880021043 Aquastar® CombiTitrant 2  one component reagents 2 mg H2O/ml  (for North America only) 1 L Glass bottle
1880051003 Aquastar® CombiTitrant 5  one component reagents 5 mg H2O/ml 1 L Glass bottle
1880052503 Aquastar® CombiTitrant 5  one component reagents 5 mg H2O/ml 2.5 L Glass bottle
1880061003 Aquastar® CombiTitrant 5 Keto  one component reagents 5 mg H2O/ml 1 L Glass bottle
1880101003 Aquastar® Titrant 5 two component reagents  5 mg H2O/ml (not for North America) 1 L Glass bottle
1880101043 Aquastar® Titrant 5 two component reagents  5 mg H2O/ml (for North America only) 1 L Glass bottle
1880102503 Aquastar® Titrant 5 two component reagents  5 mg H2O/ml (not North America) 2.5 L Glass bottle
1880102543 Aquastar® Titrant 5 two component reagents  5 mg H2O/ml (for North America only) 2.5 L Glass bottle
1880111003 Aquastar® Titrant 2 two component reagents  2 mg H2O/ml (not for North America) 1 L Glass bottle
1880111043 Aquastar® Titrant 2 two component reagents  2 mg H2O/ml (for North America only) 1 L Glass bottle
Aquastar® Certified Water Standards according to ISO 17034
1880510013 Aquastar® Water standard 0.1 % CRM (not for North America) 10 x 8 ml Ampoules
1880510313 Aquastar® Water standard 0.1 % CRM (for North America only) 10 x 8 ml Ampoules
1880520013 Aquastar® Water standard 1 % CRM (not for North America) 10 x 8 ml Ampoules
1880520313 Aquastar® Water standard 1 % CRM (for North America only)) 10 x 8 ml  Ampoules
1129390013 Aquastar® Lactose standard 5 % CRM 10 g PE bottle
1880550013 Aquastar® Oil standard CRM (15 to 30 ppm water) 10 x 8 ml Ampoules
Note: more products will follow 2020 - update 06. April. 2020