Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Reagents for Fusion
Spectromelt® Fluxes for Highest Precision
Fusion is an effective method of XRF sample preparation especially when a high degree of precision is needed. For optimal measurement accuracy, interferences caused by trace impurities in the sample are to be minimized by using a high-purity flux. Spectromelt® borates are the ideal fluxing agents for reliable XRF analysis results: Their exceptionally high purity and batch-to-batch consistency facilitates very homogeneous samples, increases measurement precision and minimizes the need for instrument adjustments.
Our portfolio comprises XRF fluxes based on lithium borates (Spectromelt® A series) as well as flux based on sodium borates (Spectromelt® B 10). All our Spectromelt® fluxing agents feature extensive specifications of trace impurities and are available as granulated material or as fused particles. The fused particles are spherical, have a small particle size distribution and a higher bulk density. They are especially easy to handle and show a low loss on ignition.
Materials
If you have trouble finding the products you're looking for, please contact our customer support for assistance.