Determination of PAHs in Paprika Powder Using a Novel SPE Cartridge
By: Katherine K. Stenerson
Principal R&D Scientist
Introduction
Paprika is a spice made from dried sweet peppers, and is used for flavor and color in many types of cuisine. Contamination with polynuclear aromatic hydrocarbons (PAHs) can occur when pepper plants are exposed to these pollutants in the environment and/or during the drying process. The use of herbs in cooking and food production has become increasingly popular, thus exposure to PAHs, specifically those with carcinogenic properties, is of concern. The European Union (EU) has gone so far as to regulate the maximum allowable levels of several PAHs in dried herbs and spices.
Specifically for paprika, the maximum allowable levels have been set to 10 ng/g for benzo[a]pyrene and 50 ng/g total for the sum of benzo[a]pyrene, chrysene, benzo[a]anthracene and benzo[b]fluoranthene.1
The sample preparation methods used in the testing of PAHs in herbs and spices require solvent extraction, followed by a cleanup step. The cleanup methods that have been used include gel permeation chromatography (GPC), and solid phase extraction (SPE) with silica gel. 2,3 In the last several years, a new approach for cleanup of high background samples to be analyzed for PAHs has utilized a dual layer SPE cartridge containing a Florisil® top layer and Z-Sep/C18 bottom layer. This cartridge, the Supelclean™ EZ-POP NP, has been used for direct extraction of PAHs from edible oils and for cleanup of extracts of biota samples.4,5 In this work, PAHs were analyzed from paprika samples using an optimized QuEChERS extraction followed by cleanup with the EZ-POP NP SPE cartridge. Compared to QuEChERS cleanup with loose sorbents, the EZ-POP NP yielded a much cleaner extract. Optimization of the extraction and EZ-POP NP cleanup procedures resulted in recoveries of >70% at a spiking level of 10 ng/g; calculated without internal standard correction.
Experiment
Paprika samples were obtained at a local grocery store. Samples for evaluation of method accuracy and reproducibility were spiked at 10 ng/g with a mixture containing the PAHs listed in Table 1. After spiking, samples were allowed to equilibrate for 1 hour prior to extraction. Sample extracts were prepared using the procedure described in Figure 1. Extracts were then subjected to SPE cleanup using the EZ-POP NP cartridge, as detailed in Figure 2. Analysis was done by GC/MS/MS using the conditions shown in Table 2 and the transitions in Table 1. An internal standard mixture was added to the extracts just prior to the GC analysis, and was used only for monitoring instrument performance. Samples were quantitated against a matrix-matched calibration curve prepared in unspiked paprika extract.
Table 1. PAHs Analyzed and Transitions Used for GC/ MS/MS Analysis
Peak No. Compound RT MRM 1 CE MRM 2 CE
1 Naphthalene-d8 (IS)   5.21 136/108 20 136/84 25
2 Naphthalene   5.25 128/102 20 128/78 20
3 Acenaphthylene   7.62 152/126 30 152/102 30
4 Acenaphthene   7.88 153/127 30 152/126 30
5 Fluorene   9.2 166/165 15 165/164 20
6 Phenanthrene 13.5 178/152 25 176/150 25
7 Anthracene 13.71 178/151 30 176/150 25
8 Fluoranthene-d10 (IS) 20.76 212/208 35 210/208 15
9 Fluoranthene 20.91 202/152 30 201/200 15
10 Pyrene 22.95 201/200 15 200/174 25
11 Benz[a]anthracene 29.42 228/226 30 114/101 10
12 Cyclopenta[cd]pyrene 29.72 226/225 30 226/224 40
13 Chrysene 29.81 228/226 30 113/112 10
14 5-Methyl chrysene (IS) 31.7 242/241 20 241/239 30
15 Benzo[b]fluoranthene 33.60 252/250 30 126/113 10
16 Benzo[k]fluoranthene 33.67 252/250 30 126/113 10
17 Benzo[j]fluoranthene 33.71 252/250 30 126/113 10
18 Benzo[a]pyrene 34.64 252/250 35 125/124 10
19 Perylene-d12 34.84 264/260 35 130/116 15
20 Dibenz[a,h]anthracene 37.13 278/276 35 125/124 10
21 Indeno[1,2,3-cd] pyrene 37.2 138/137 10 137/136 15
22 Benzo[g,h,i]perylene 38.13 138/137 15 137/136 15
23 Dibenzo[a,l]pyrene 41.77 302/301 20 302/300 40
24 Dibenzo[a,e]pyrene 43.27 302/301 40 302/300 40
25 Coronene (IS) 43.76 300/299 35 300/298 50
26 Dibenzo[a,i]pyrene 44.29 302/301 40 302/300 40
27 Dibenzo[a,h]pyrene 44.68 302/301 20 302/300 40
Results & Discussion
GC/MS/MS Conditions. The analysis conditions were optimized for PAHs. Higher injection port and MS temperatures, along with a  pressure  pulsed  injection and a 2 mm ID inlet liner,  were used to improve   response and peak shape. The oven temperature program was based on that used by  Anderson  et  al., and was necessary to provide  chromatographic resolution of isomers such as benzo[b], [k] and [j] fluoranthene, that cannot be distinguished by their MRM transitions.6 This temperature program, in combination with the GC capillary column (SLB®-PAHms), provided adequate resolution of all isomeric sets. An example of this is shown in Figure 3 for the isomers benzo [b], [k] and [j] fluoranthene. These isomers cannot be separated on a 5% phenylmethylsilicone or “X-5” type phase.
Sample extracts were prepared using the procedure described
Figure 1.Sample Extraction Procedure Used for PAHs from Paprika
Cleanup Procedure Using EZ-POP NP SPE
Figure 2. Cleanup Procedure Using EZ-POP NP SPE for Paprika Extract
Adequate resolution of all isomeric sets
Figure 3. GC/MS/MS Analysis of Benzo [b], [k], and [j] Fluoranthenes on the SLB®-PAH MS Columns; 10 ppb in Paprika Extract
Effect of Extraction Solvent on Recovery
Figure 4. Effect of Extraction Solvent on Recovery of 5 & 6 Ring PAHs from Spiked Paprika (10 ng/g)
Table 2. GC Analysis Conditions
column SLB®-PAHms, 30 m x 0.25 mm I.D., 0.25 µm (28340-U)
oven 60 °C (1 min), 40 °C/min to 180 °C, 3 °C/min to 230 °C, 1.5 °C/min to 235 °C (10 min), 15 °C/min to 280 °C (3 min), 15 °C/min to 350 °C (15 min)
inj. temp 300 °C
carrier gas helium, 0.8 mL/min, constant
detector MRM, per Table 1
MS source temp. 250 °C
Quadrupole temps 200 °C (Q1 & Q2)
injection 1 µL, pulsed splitless (60 psi until 0.75 min, splitter on at 0.75 min)
Optimization of Sample Preparation Procedure
Sample extraction was done based on a “QuEChERS-  like” approach. The composition of the extraction solvent was optimized from the 100% acetonitrile normally used  for QuEChERS to increase recovery of the heavier PAHs.
Figure 4 presents a comparison of absolute response for the heavier 5 & 6 ring PAHs obtained from paprika spiked at 10 ng/g, and extracted with 100% acetonitrile and a mixture of 90:10 acetonitrile:methylene chloride. The resulting extracts were analyzed before SPE cleanup to evaluate the effect of the extraction solvent only.
The 5&6 ring PAHs showed better recovery using the 90:10 mixture, as indicated by the higher responses. As expected, the extract produced using the 90:10 mixture showed more background, visible as oily residue. For this reason, higher percentages of methylene chloride in the extraction solvent were not evaluated.
For cleanup, the EZ-POP NP SPE cartridge provided better cleanup than QuEChERS. The EZ-POP NP cartridge was originally developed for analysis of PAHs from edible oils, specifically olive oil. Acetonitrile is the elution solvent recommended for use with edible oil samples, as it provides a balance between analyte recovery and background retention. Previous work showed that the addition of methylene chloride increased PAH recovery; but it had the unwanted side effect of increasing the amount of background in the final extract.4 However, since the solvent extracts of the paprika contained substantially less oil, it was possible to modify the cleanup procedure to increase recoveries of the 5 and 6 ring  PAHs without increasing background to an unacceptable level. Specifically, the composition of the elution solvent was changed to include methylene chloride. Figure 5 shows a comparison of absolute response obtained for 4-6 ring PAHs  using  different  elution  solvents  during the EZ-POP NP cleanup. The samples processed were paprika (10 ng/g spiking level) extracted using 90:10 acetonitrile:methylene chloride. The responses of all PAHs increased with the addition of methylene chloride to the elution solvent. The difference in responses between 10 and 20% addition was minimal for some compounds; however, the background in the final extract did increase. Thus, 90:10 acetonitrile:methylene chloride was chosen as the elution solvent to be used in the EZ-POP NP cleanup.
A comparison of absolute response
Figure 5. Effect of Elution Solvent Composition Used During EZ-POP NP Cleanup on Recovery of 4-6 Ring PAHs from Spiked Paprika (10 ng/g)
Paprika Extracts Cleaned Using EZ-POP NP SPE
Figure 6. Paprika Extracts Cleaned Using EZ-POP NP SPE and QuEChERS with Similar Sorbents. Z-Sep, Z-Sep/C18 and Z-Sep+ are QuEChERS cleanups
Using GC/MS-scan for comparison
Figure 7. GC/MS Scan Comparison of Paprika Extracts; EZ-POP NP, QuEChERS with Z-Sep+, and No Cleanup.
GC/MS/MS Analysis of PAHs in Paprika
Figure 8. GC/MS/MS Analysis of PAHs in Paprika, Spiked at 10 ng/g, After Cleanup Using EZ-POP NP. Peak IDs are as indicated in Table 1.
Background Reduction
In the case of dry samples such as paprika, QuEChERS cleanup using loose sorbents does not provide enough capacity for reducing background to an acceptable level. If analysis is for pesticides, dual layer SPE cartridges containing carbon and primary secondary amine (PSA) or aminopropyl functionalized silica can be used for cleanup. However, for PAHs, carbon cannot be used as it will strongly retain these compounds, resulting in poor recovery. EZ-POP NP does not contain carbon, and thus is suitable for PAHs. Its cleanup capacity was compared directly to QuEChERS using blends containing similar sorbents to those in the EZ-POP NP cartridge. Extracts of paprika generated using 90:10 acetonitrile:methylene chloride and cleaned using EZ-POP NP and QuEChERS are shown in Figure 6. Compared to no cleanup, both approaches reduced color, with the EZ-POP NP having the lightest color. The sample cleaned with Z-Sep showed some oily residue, as seen on the inside of the vial. Using GC/MS-scan for comparison (Figure 7), the EZ-POP NP cleanup produced the lowest background, especially with regard to removal of C16 and C18 fatty acids. Only Z-Sep+ data is shown here, as it had the lowest background of the three QuEChERS sorbents included in the comparison. Using the final, optimized cleanup method with EZ-POP NP, all PAHs were detected free of background as shown in Figure 8.
PAH Spike Recoveries and Analysis of Unknown Samples
Recovery and reproducibility data for analyses of paprika samples spiked at 10 ng/g is presented in  Table 3. All recovery values were determined without internal standard correction and thus are absolute. Overall, recoveries were very good (>70%) with RSD values of <15%. The exceptions were fluorene and dibenzo[a,h] pyrene. There was some matrix interference eluting with the fluorene peak (present in both MRMs monitored), and for dibenzo[a,h]pyrene, response was erratic from the spikes, which resulted in poor reproducibility.
Table 3. Recovery and Reproducibility for PAHs from Paprika, Spiking Level of 10 ng/g (n = 3)
n=3 avg. % Recovery % RSD
Naphthalene 124% 11%
Acenaphthene   78% 13%
Acenaphthylene   76% 15%
Fluorene   52% 27%
Phenanthrene 102%   4%
Anthracene   78% 12%
Fluoranthene   95%   7%
Pyrene 100%   3%
Benz[a]anthracene   99%   9%
Cyclopenta[c,d]pyrene   70% 12%
Chrysene   99%   6%
5-Methyl chrysene   91% 10%
Benzo[b]fluoranthene   89%   6%
Benzo[k]fluoranthene   82%   6%
Benzo[j]fluoranthene   97%   8%
Benzo[a]pyrene   91% 12%
Dibenz[a,h]anthracene   84%   6%
Indeno[1,2,3-cd]pyrene   86% 12%
Benzo[g,h,i]perylene   80%   5%
Dibenzo[a,l]pyrene 108% 12%
Dibenzo[a,e]pyrene   82%   5%
Dibenzo[a,i]pyrene 108% 15%
Dibenzo[a,h]pyrene 147% 36%
In addition to the paprika used for spike recovery, two more brands were analyzed unspiked. One was labeled as “smoked paprika” and thus was expected to contain substantially higher PAH levels. Both brands were analyzed in duplicate, and the results are summarized in Table 4. Several PAHs were detected in brand A; but none were at levels above the EU limits for herbs and spices. The smoked sample, on the other hand, contained detectable levels of all four of the PAHs listed in the EU regulation. Benzo[a]pyrene was present below the maximum allowable level of 10 ng/g; however, the combined levels of this PAH plus benzo[a] anthracene, chrysene and benzo[b]fluoranthene were well above the 50 ng/g maximum. This is to be expected, since the smoking process produces PAHs which subsequently end up in the paprika. However, the EU regulation specifically states that since smoked herbs are generally consumed in such small quantities, they are exempt from these regulatory limits.
Table 4. Analysis of Paprika in Unspiked Samples (% RPD = % reproducibility)
  Brand A Brand B*
n=2 Conc. (ng/g) %RPD Conc. (ng/g) %RPD
Naphthalene 69.8 5%   302   3%
Acenaphthene   3.0 10%   211.6   1%
Acenaphthylene   ND       27.7   4%
Fluorene   ND     272.2   6%
Phenanthrene 25.1 2% 2833 56%
Anthracene   ND     677   4%
Fluoranthene   ND     995   8%
Pyrene   ND     884   7%
Benz[a]anthracene   ND     177.6   7%
Cyclopenta[c,d]pyrene   7.1 1%     99   7%
Chrysene   ND     103.1   8%
5-Methyl chrysene   ND        0.6 25%
Benzo[b]fluoranthene   ND       10.8   5%
Benzo[k]fluoranthene   ND        ND  
Benzo[j]fluoranthene   ND         6.4   5%
Benzo[a]pyrene   ND         8.3   6%
Dibenz[a,h]anthracene   ND         ND
Indeno[1,2,3-cd]pyrene   ND         2.3   2%
Benzo[g,h,i]perylene   ND         2.6   5%
Dibenzo[a,l]pyrene   ND         ND  
Dibenzo[a,e]pyrene   ND         ND  
Dibenzo[a,i]pyrene   ND         ND  
Dibenzo[a,h]pyrene   ND         ND  
*smoked paprika
Heavier PAHs can be difficult to analyze at low levels by GC/MS due to issues with response and peak shape. Instrument performance can rapidly decline with the injection of dirty samples. To determine if this was occurring with analysis of the paprika extracts, internal standards of non-target PAHs from 4-6 rings were added to the extracts prior to analysis. Response of these compounds was then monitored throughout the run. No decline in response was observed for any of the internal standards, and the variation in area counts throughout the run was <15% for each. This indicates that the cleanup was adequate to produce a rugged GC method.
Conclusions
A sample preparation method was developed for the low level analysis of PAHs in paprika powder. The extraction protocol utilized a “modified QuEChERS” approach in which methylene chloride was added to the extraction solvent to increase recoveries. Cleanup was achieved using SPE with the EZ-POP NP cartridge with the addition of methylene chloride to the acetonitrile elution solvent. This modification resulted in absolute recoveries of the 4-6 ring PAHs of >80% with RSD values < 15% for most. The SPE cleanup reduced background in the extract, especially for C16 and C18 fatty acids, significantly compared to QuEChERS cleanup using loose sorbents. Analysis of the PAHs on an application specific GC column (the SLB®-PAHms) was necessary to chromatographically resolve isomeric sets which cannot be discerned based on MRMs.
References
  1. Commission Regulation (EU) 2015/1933 of 27 October 2015 amending Regulation EC/1881/2006 as regards maximum levels for polycyclic aromatic hydrocarbons in cocoa fibre, banana chips, food supplements, dried herbs and dried spices. Off. J. Eur. Union 2015, L282/11.
  2. Rozentale, I.; Lun, A. Y.; Zacs, D.; Bartkevics, V., The occurrence of polycyclic aromatic hydrocarbons in dried herbs and spices. Food Control. 2018, 83, 45-53.
  3. Monado-Maraña, O.; Pérez, R.L.; Escandar, G.M.; Muñoz de Peña, A.; Galeano-Díaz, T., Combination of liquid chromatography with multivariate curve resolution-alternating least- squares (MCR-ALS) in the quantitation of polynuclear aromatic hydrocarbons present in paprika samples. J. Agric. Food Chem. 2016, 64, 8254-8262.
  4. Stenerson, K.K.; Shimelis, O.; Halpenny, M.; Espenschied, K.; Ye, M.M, Analysis of polynuclear aromatic hydrocarbons in olive oil after solid-phase extraction using a dual-layer sorbent cartridge followed by high-performance liquid chromatography with fluorescence detection. J. Agric. Food Chem. 2015, 63, 4933-4939.
  5. Cabillic, J.; Fallot, C.,Final Report on the Feasibility Study of QuEChERS Extraction and / or Purification for Analysis of PAHs in Biota, AQUAREF Report 2015 - 22 p.
  6. Anderson, K.A.; Szelewski, M.J.; Wilson, G; Quimby, B.D.; Hoffman, P.D., Modified ion source triple quadrupole mass spectrometer gas chromatograph for polycyclic aromatic hydrocarbon analyses. J. Chrom. A. 2015, 1419, 89-98.
For the complete range of our PAH standards, please visit SigmaAldrich.com/pahstandards
Materials