[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Thin Layer Chromatography](https://www.sigmaaldrich.com/US/en/applications/analytical-chemistry/thin-layer-chromatography)High-Performance Thin-Layer Chromatography: A Fast and Efficient Fingerprint Analysis Method for Medicinal Plants
# High-Performance Thin-Layer Chromatography: A Fast and Efficient Fingerprint Analysis Method for Medicinal Plants
__Tiên Do, Melanie Broszat, Matthias Nold__
[Article from Analytix Reporter - Issue 2](https://www.sigmaaldrich.com/US/en/collections/analytix-reporter)
Ginsenosides are triterpene saponins. Most ginsenosides are composed of a dammarane skeleton (17 carbons in a four-ring structure) with various sugar moieties (e.g. glucose, rhamnose, xylose and arabinose) attached to the C- 3 and C-20 positions.
Over 30 ginsenosides have been identified and classified into two categories:
• 20(S)- protopanaxadiol (PPD) (Rb1, Rb2, Rb3, Rc, Rd, Rg3, Rh2, Rs1)
• 20(S)-protopanaxatriol (PPT) (Re, Rf, Rg1, Rg2, Rh1). The difference between PPTs
and PPDs is the presence of a carboxyl group at the C-6 position in PPDs.
__Figure 1.__
Moreover, several rare ginsenosides, such as the ocotillol saponin F11 (24-R-pseudoginsenoside) and the pentacyclic oleanane saponin Ro (3,28-o-bisdesmoside) have also been identified. Asian ginseng (Panax ginseng) is commonly known as the true ginseng.
In this article, HPTLC methods suitable for the analysis of ginsenosides are presented, using CAMAG equipment, MilliporeSigma TLC plates, analytical standards and extract reference materials. The extract reference materials are manufactured by HWI Analytik and exclusively distributed by MilliporeSigma.
Detection of ginsenosides in the HPTLC fingerprint of different Panax species (roots and root extracts) is obtained by following the HPTLC methods of the Ph. Eur. monograph,1 USP DSC 2015 monograph2 and the method of the HPTLC Association (International Association for the Advancement of HPTLC)3 by comparison of the RF values and colors of reference substances and matching zones in the root extract. Depending on the regulation followed, one of the three methods of identification can be used.
## Recommended CAMAG Devices:
Automatic TLC Sampler (ATS 4), Automatic Developing Chamber (ADC 2), TLC Visualizer 2, Chromatogram Immersion Device 3, TLC Plate Heater 3, and visionCATS
## Derivatization Reagent:
Anisaldehyde1 or sulfuric acid reagent2,3
## Sample:
0.015 g/mL extract (HWI extract) in 70% methanol1 *Note: Deviation from methods 2 and 3 for the sample preparation*
## Standards:
Standard solutions of ginsenosides were prepared in a concentration of 0.5 mg/mL in methanol.
*Note: Deviation from method 3 for the application volumes*
## Chromatography Following USP <203> 4:
__Stationary phase:__ HPTLC Si 60 F254 20 x 10 cm (MilliporeSigma)
__Sample application:__ 4 µL each of test solution and 2 µL of standards are applied as 8 mm bands, 8 mm from lower edge, 20 mm from the left edge1, 3
*Note: Deviation from method 3 for the sample preparation and the application volume of reference and test solutions*
4 µL each of test solution and 4 µL of standards are applied as 8 mm bands, 8 mm from lower edge, 20 mm from the left edge2
*Note: Deviation from method 2 for the sample preparation*
__Developing solvent:__ Ethyl acetate – water – butanol 25:50:100 (*v/v/v*) - upper layer1
Dichloromethane – ethanol – water 70:45:6.5 (*v/v/v*)2
Chloroform – ethyl acetate – methanol – water 15:40:22:9 (*v/v/v/v*)3
__Development:__ In the ADC 2 with an unsaturated chamber and after conditioning at 33% relative humidity for 10 min using a saturated solution of magnesium chloride1
Development is performed with ADC 2, saturated for 20 minutes with the developing solvent (filter paper). Prior to the development the plate is conditioned for 10 min to a relative humidity of 33% (with a saturated solution of MgCl2).2, 3
__Developing distance:__ 70 mm (from lower edge)1, 2; 80 mm (from lower edge)3
__Plate drying:__ 5 min in a stream of cold air
__Derivatization:__ The plate is immersed (immersion speed: 3 cm/s, immersion time: 0 s) into anisaldehyde reagent (mixture of 0.5 mL of *p*-anisaldehyde, 10 mL of glacial acetic acid, 85 mL of methanol, and 5 mL of sulfuric acid) with the Chromatogram Immersion Device 3 and heated for 5 min at 105°C.1
The plate is immersed (immersion speed: 3 cm/s, immersion time: 0 s) into sulfuric acid reagent (10% in methanol) with the Chromatogram Immersion Device 3 and heated for 5 min at 100 °C.2, 3
__Evaluation:__ Documentation under method 3 is with 100 °C white light1, 2, 3 and UV 366 nm2, 3 after derivatization with the TLC Visualizer 2
## Results and Discussion:
### HPTLC chromatograms of ginsenoside standards and a *Panax ginseng* root extract
HPTLC chromatograms after derivatization. Tracks
1-17: ginsenosides, track 18: protopanaxadiol, track 19: *Panax ginseng* root extract (article no.: 05115001 batch: HWI01294)
__Method 1:__According to the Ph. Eur.
HPTLC chromatograms under white light after derivatization
__Method 2:__According to the Tienshi ginseng method from
HPTLC chromatograms under UV 366 nm and under white light after derivatization
__Method 3:__According to the HPTLC Association
HPTLC chromatograms under UV 366 nm and under white light after derivatization.
## HPTLC chromatograms of plants containing ginsenosides (different Panax species)
*P. ginseng, P. quinquefolium, P. notoginseng, P. japonicus, P. vietnamensis* roots and root extracts were collected and analyzed. The *P. ginseng* root extract (article no.: 0511-50-01 batch: HWI01294) was used as botanical reference material to identify Asian ginseng (the ginsenoside Rf should be present and F11 absent).
__Method 1:__ According to the Ph. Eur.
HPTLC chromatogram under white light after derivatization.
__Track 1:__ ginsenoside Rf (green arrow); __2:__ ginsenoside F11 (red arrow); __3:__ *P. ginseng* root extract (article no.: 0511-50-01 batch: HWI01294, ginsenoside Rf highlighted with the green arrow); __4:__ P. ginseng root (ginsenoside Rf highlighted with the green arrow); __5:__ *P. quinquefolium* root extract (American ginseng, ginsenoside F11 highlighted with the red arrow);
__6:__ *P quinquefolium* root (American ginseng, ginsenoside F11 highlighted with the red arrow); __7:__ *P. notoginseng* root extract;
__8:__ *P. notoginseng* root; __9:__ *P. japonicas* root; __10:__ *P. vietnamensis* root; __11:__ wild Vietnamese ginseng root.
__Method 2:__According to the Tienshi ginseng method from
HPTLC chromatograms under UV 366 nm and under white light after derivatization
__Method 3:__According to the HPTLC Association
HPTLC chromatograms under UV 366 nm and under white light after derivatization.
## Method 2 & 3 Parameter:
HPTLC chromatograms under white light and UV 366 nm after derivatization with ginsenoside Rf (green arrows) and ginsenoside F11 (red arrows).
__Track 1: P. ginseng__ root extract (article no.: 05115001 batch: HWI01294, ginsenoside Rf highlighted with the green arrows); __2:__ *P. ginseng* root (ginsenoside Rf highlighted with the green arrows); __3: P. quinquefolium__ root extract (American ginseng, ginsenoside F11 highlighted with the red arrows); __4:__ *P quinquefolium* root (American ginseng, ginsenoside F11 highlighted with the red arrows); __5: P. notoginseng__ root extract; __6:__ *P. notoginseng* root; __7:__ *P. japonicas* root; __8:__ *P. vietnamensis* root;
__9:__ wild Vietnamese ginseng root.
All shown methodologies are suitable for detection of ginsenosides in different Panax species. Ginsenoside Rf is unique to Asian ginseng while F11 is found exclusively in American ginseng. Thus the Rf/F11 ratio is used as a phytochemical marker to distinguish American ginseng from Asian ginseng. In the botanical reference material used (*Panax ginseng* root extract, article no.: 0511-50-01) the presence of Rf and absence of F11 could be confirmed with all methods and it is therefore suited for identification of Asian ginseng.
Methods 1 and 2 have the advantage in that they are in accordance with official monographs in the pharmacopoeias (Ph. Eur. resp. USP). Method 3 is an alternative method provided by the HPTLC Association. The method is improved for the separation of Rf and F11 to better distinguish American and Asian ginseng. The derivatization with sulfuric acid reagent (methods 2 and 3) leads to different colored zones under UV 366 nm, useful for identification.
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
### References
1\.
2008\. Ginseng root: Monograph in Ph. Eur. 8.0. European Directorate for the Quality of Medicines and HealthCare.. 01. Strasbourg: France.
2\.
2017\. Tienchi Ginseng Root and Rhizome Dry Extract: Monograph in USP 40-NF35. United States Pharmacopeial Convention. USA: Rockville, MD.
3\.
2016\. HPTLC identification method for Panax ginseng, HPTLC Association (www.hptlc-association.org).
4\.
2016\. High-Performance Thin-Layer Chromatography Procedure for Identification of Articles of Botanical Origin in USP 39-NF34. United States Pharmacopeial Convention.. USA: Rockville, MD.
__Related Articles__
- [Analysis of Catechins in Green Tea by TLC](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/analysis-of-catechins-in-green-tea-by-tlc)
- [High Performance Thin Layer Chromatography Plates](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/high-performance-thin-layer-chromatography-plates)
- [Identification of fixed oils by Thin layer chromatography](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/identification-of-fixed-oils-by-thin-layer-chromatography)
- [Identification of Glycyrrhizic Acid in Licorice](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/identification-of-glycyrrhizic-acid-in-licorice)
- [Tips & Tricks for Thin-Layer Chromatography](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/tips-n-tricks-for-thin-layer-chromatography)
- [History of Thin-Layer Chromatography](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/tlc-history)
- [Determination of Stevioside and Rebaudioside A in Stevia leaves, beverages and food additives using HPTLC-MS](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/tlc-ms-determination-of-stevioside-and-rebaudioside)
- [Quantification of Bitter Acids in Hops by Thin Layer Chromatography](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/tlc-ms-quantification-of-bitter-acids-in-hops)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Phytochemical Standards](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/reference-materials/phytochemical-standards)
- [Food & Cosmetic Component Standards](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/reference-materials/food-and-cosmetic-component-standards)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
An unknown error has occured.