New CRM Solutions for Paralytic Shellfish Toxins
Matthias Nold  Global Product Manager Reference Materials
Paralytic Shellfish Toxins (PSTs)Paralytic Shellfish Toxins (PSTs) are a group of highly toxic, naturally occurring alkaloids produced by certain species of marine algae. During harmful algae blooms, these toxins can accumulate in bivalve mollusks and cause severe poisoning after consumption. The occurrence of PSTs in seafood, therefore, needs to be monitored. We are introducing a series of native and isotope labeled Certified Reference Material (CRM) solutions for paralytic shellfish toxins, suitable for calibration of LC-MS / LC-IDMS (isotope dilution MS) food testing methods.
The CRM solutions are manufactured in-house under our ISO/IEC 17025 and ISO 17034 accreditation, and feature:
  • Certification by qNMR or IDMS with traceability to SI unit via NIST or NRC primary standards
  • Solutions produced gravimetrically
  • Homogeneity, accelerated and long-term stability tested by LC-MS
  • A comprehensive certificate including the overall uncertainty
As a first set of products, we are introducing the PSTs Neosaxitoxin and Saxitoxin dihydrochloride as CRM solutions both in native and 15N labeled form.
Materials