Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Maize GMO Standards
By: Kurt Vorburger, AnalytiX Lab Info 4
NK603, GA21 and CBH-3511 Starlink® ......... The use of genetically modified organisms (GMO) is still a widely debated topic in Europe. Consumers in the European Union are wary of GMOs and indeed many retailers avoid stocking foods containing them.
However, in order to measure the presence or absence of GMOs a suitable standard is required. We offer a range of GMO Maize Standards, extracted from actual plant material. It is now easier to obtain positive controls for NK603, GA21 and CBH-3511 Starlink maize varieties.
Introduction
The use of genetically modified organisms (GMO) is still a widely debated topic in Europe. Consumers in the European Union are wary of GMOs and indeed many retailers avoid stocking foods containing them.
In April 2004 the legislation regarding the labelling of genetically modified foods and feed products changed. This legislation requires that even highly modified foods such as lecithin, starch and oil, as well as feed products containing GMOs are labelled clearly as such.
Maize is now, therefore, subject to the labelling legislation, requiring more stringent control and labelling than was previously required.
However, in order to measure the presence or absence of GMOs a suitable standard is required, which has historically been difficult to source.
This range of GMO Maize Standards, contains some varieties that are not approved in the European Union and many other countries worldwide. It is now easier to obtain positive controls for three different GMO maize varieties.
back to top 
Description
These GMO standards NK603, GA21 and CBH-3511 Starlink® maize DNA, extracted from actual plant material. Total maize DNA is comprised of approximately 1% of GMO DNA. This GMO DNA was ex-tracted as described in the official protocols1 and tested in several validation studies2-4. The extracted DNA was quantified and diluted to form a 1% solution, which was then aliquoted and lyophilised, enabling a longer storage period.
There are numerous benefits to be gained from the use of these standards:
  • The process of DNA extraction for use as a positive control can be avoided, thereby eliminating the need for time-consuming manual extraction procedures.
  • Since the DNA is extracted from plant material, the application of these standards is no longer limited to a certain sequence information or protocol. Therefore, it is possible to use them with all DNA–based methods and even with your own optimized protocols
  • The world wide availability of GMO standards having the same characteristics and composition can ensure the standardization of measurements and procedures, therefore allowing comparison of results obtained in different facilities.
back to top 
Use
The GMO standards are used solely as qualitative standards. The DNA is lyophilised and can be used as a positive control after resolution. The native maize content of the samples is approximately 100 times higher than GMO maize.
Find a complete listing of all GMO standards in our product catalog.
back to top 
References
  1. SLMB2001: Swiss Food Manual (SLMB 2001) Eidgenössische Drucksachen und Materialzentrale CH-3003 Bern, Switzerland (2001)
  2. U. Pauli, B. Schouwey, P. Hübner, P. Brodmann, A. Eugster. Quantitative Detection of Genetically Modified Soybean and Maize: Method Evaluation in a Swiss Ring Trial. Mitt. Gebiete Lebensm. Hyg. 92, 145-158 (2001)
  3. M. Lipp, A. Bluth, F. Eyquem, L. Kruse, H. Schimmel, G. Van den Eede, E. Anklam. Validation of a method based on polymerase chain reaction for the detection of genetically modified organisms in various processed foodstuffs. Eur Food Res Technol 212, 497–504 (2001)
  4. H. Hird, J. Powell, M.-L. Johnson, S. Oehlschlage.: Determination of Percentage of RoundUp Ready Soya in Soya Flour Using Real-Time Polymerase Chain Reaction: Interlaboratory Study. Journal of AOAC International 86, 66–71 (2003)
Materials