Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
CERTAN® Capillary Vials
By: Nicole Amann, Analytix Volume 5 Article 6
The advantages of a sealed ampul with the flexibility of a screw cap bottle or a septum vial. Ideal for the storage of volatile samples and reference standards.
By Nicole Amann, Product Manager Analytical Standards
Features
  • 1.5 mL, 4.5 mL and 10 mL volumes
  • Amber glass
  • 1.2 mm I.D. x 28 mm height capillary opening permits sample withdrawal or addition via syringe needle
  • PTFE-lined screw-cap gives a sure seal
back to top
Advantages
  • No appreciable change in concentration, even when open
  • Secure storage with minimal evaporation
  • Minimizes contamination risk
  • Sample filling and removal using a standard GC syringe
  • Essentially spill-proof
back to top
CERTAN® Vials are intended for these and other important analytical applications
  • Storage of liquids removed from conventional glass ampuls
  • Storage of stock and standard solutions
  • Storage of difficult samples such as volatile halogen compounds or BTEX aromatics
  • Archiving of production or QC samples and extracts
back to top