[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Bacteriology](https://www.sigmaaldrich.com/US/en/applications/clinical-testing-and-diagnostics-manufacturing/bacteriology)Microbial Detection Using Chromogenic and Fluorogenic Culture Media
# Microbial Detection Using Chromogenic and Fluorogenic Culture Media
As the rapid detection and identification of microorganisms becomes increasingly vital in microbiology, fluorogenic and chromogenic substrates have emerged as powerful tools, leveraging the specific enzymatic activities of certain microorganisms. By incorporating synthetic fluorogenic or chromogenic substrates into primary selective media, enumeration and detection can be conducted directly on the isolation plate.
Chromogenic and fluorogenic media are specialized microbiological growth media that contain enzyme substrates linked to a chromogen (color reaction), fluorogen (light reaction) or a combination of both. These media offer significant advantages for the enumeration, detection, and identification of microorganisms. They facilitate the screening of all major food pathogens, including *Salmonella, Campylobacter, Listeria, Listeria monocytogenes, Staphylococcus/S. aureus,* Coliforms, *E. coli* as well as specific target microorganisms such as, *E. coli* O157.
### Section Overview
- [Principle of the Fluorogenic and Chromogenic Technique](https://www.sigmaaldrich.com#fluorogenic-chromogenic-technique)
- [Advantages of Chromogenic Media](https://www.sigmaaldrich.com#chromogenic-media)
- [ChromoCult® Coliform Agar](https://www.sigmaaldrich.com#coliform-agar)
- [ChromoCult® Coliform Agar ES](https://www.sigmaaldrich.com#coliform-agar-es)
- [ReadyCult® Fluorogenic Media](https://www.sigmaaldrich.com#fluorogenic-media)
- [ReadyCult® Coliforms 100](https://www.sigmaaldrich.com#coliforms-100)
- [ReadyCult® Enterococci 100](https://www.sigmaaldrich.com#enterococci-10)
- [MilliChrome™ Chromogenic Culture Media](https://www.sigmaaldrich.com#chromogenic-culture-media)
- [Related Products](https://www.sigmaaldrich.com#related-products)
* * *
## Principle of the Fluorogenic and Chromogenic Technique[](https://www.sigmaaldrich.com)
The comparison between traditional and enhanced media formulations containing chromogenic substrates is a significant topic in microbiology today. Fluorogenic enzyme substrates typically consist of a specific substrate tailored for a particular enzyme (e.g., sugar or amino acid) and a fluorogen, such as 4-methylumbelliferone, which emits a blue fluorescence in a positive result. The substrate MUG (4-methylumbelliferyl-β-D-glucuronide) is commonly added to traditional media for the detection of *Escherichia coli* or *E. coli O157.*
Chromogenic substrates such as ONPG, X-Gal, or X-Glu, combined with a specific medium selectivity, facilitate the easy differentiation of microorganisms based on the coloration of grown bacteria. The enzyme characteristic of the target organism cleaves the chromogenic substrate into a sugar component and a chromogen, resulting in a distinct color change in liquid broth or on agar plates in the presence of oxygen. This enables direct confirmation of the target organism without the need for further testing.
Furthermore, it is now possible to detect and differentiate multiple organisms on a single plate. By integrating several chromogenic substrates with appropriate selectivity, different species or groups of microorganisms can be differentiated on one plate. Most chromogenic media are both selective and differential, inhibiting non-target organisms while allowing target pathogens to develop colored colonies due to their metabolism of one or more chromogenic enzyme substrates.
## Chromogenic substrates and selective agents
| | | | |
|------------------------------------------------------|----------------------------------------------------------------|-----------------------------------------------------------------------------|--------------------------------------------------------------------------------------------|
| __Species__ | __Enzyme__ | __Substrate__ | __Selective Agents__ |
| *Bacillus cereus* | β-glucosidase, Phosphatidylinositol-specific Phospholipase C | indoxyl-β-glucopyranoside, indoxyl-myo-inositol-1-phosphate | polymyxin B |
| *Campylobacter* | na | na | deoxycholate, cefoperazone, amphotericin B |
| *Candida* | β-acetylgalactosaminidase, alkaline phosphatase | indoxyl-N-acetyl-β-D-glucosaminide, indoxyl-phosphate | Chloramphenicol, Gentamicin |
| *Clostridium perfingens* | β-glucosidase (plus sucrose fermentation) | indoxyl-β-D-glucoside | D-cycloserine, polymixin B |
| Coliforms/*E. coli* | β-glucuronidase, β-galactosidase | indoxyl-β-glucuronide, Indoxyl-β-galactoside | bile salts, tergitol 7<R>, SDS, novobiocin, cefsulodin |
| *Cronobacter (E.sakazakii)* | a-glucosidase | indoxyl-a-D-glucoside | deoxycholate, crystal violet, sodium thiosulfate |
| *E. coli O157* | β-glucosidase, a-galactosidase | indoxyl-β-D-glucuronide, indoxyl-a-galatoside | bile salts, SDS, crystal violet, potassium tellurite, novobiocin, cefixime |
| Enterococci | β-D-glucosidase | indoxyl-β-glucoside | sodium azide, polysorbate 80 |
| Extended Spectrum ß-Lactamase Enterobacteria (ESBL) | β-D-glucosidase | indoxyl-β-glucoside | cefpodoxime, cefotaxime, ceftazidime |
| *Klebsiella* | β-D-ribofuranosidase, β-D-glucosidase | indoxyl-β-D-ribofuranoside, indoxyl-β-D-glucoside | bile salts, SDS, carbenicillin |
| *Listeria spp.* | β- glucosidase | indoxyl-β-glucoside | lithium chloride, ceftazidime, amphotericin B, nalidixic acid, polymyxin B |
| *L. monocytogenes* | Phosphatidylinositol-specific Phospholipase C, β- glucosidase, | indoxyl-β-glucoside, indoxyl-myo-inositol-1-phosphate | lithium chloride, ceftazidime, amphotericin B, nalidixic acid, polymyxin B |
| *Pseudomonas* | β-Alanyl arylamidase | 7-Amido-1-pentyl-phenoxazin-3-one | cetrimide |
| *Salmonella* | a-galactosidase, lipase | indoxyl-a-galactoside, indoxyl-fatty acid ester | sodium deoxycholate |
| MRSA (Methicillin Resistant *Staphylococcus aureus*) | a-glucosidase | indoxyl-a-D-glucopyranoside | methicillin, high concentration of sodium chloride |
| *Staphylococcus aureus* | a-glucosidase, phosphatase, deoxyribonuclease | indoxyl-a,D-glucoside, phenolphthalein phosphate, indoxyl-phosphate | tellurite, lithium chloride |
| Streptococci | β-glucuronidase | indoxyl-β-glucuronide | sodium azide |
| UTI (Urinary Tract Infections) | β-glucosidase, β-galactosidase | indoxyl-β-glucopyranoside, indoxyl-β-galactoside | \- |
| *Vibrio* | β-glucosidase, β-galactosidase | indoxyl-β-glucoside, indoxyl-β-galactoside | high concentration of sodium chloride, sodium thiosulphate, sodium citrate, sodium cholate |
| VRE (Vancomycin Resistant Enterococci) | a-glucosidase, β-glucosidase, β-galactosidase | indoxyl-a-glucopyranoside, indoxyl-β-glucopyranoside, indoxyl-β-galactoside | vancomycin |
| Yeasts and Molds | β-N-acetylgalactosaminidase, β-xylosidase | indoxyl-N-acetyl-β-D-glucosaminide, indoxyl-β-D-xyloside | oxytetracycline |
In Table 1,known substrates and selective agents are listed and give some idea about additional possibilities.
* * *
## Advantages of Chromogenic Media[](https://www.sigmaaldrich.com)
Chromogenic media are invaluable media for the detection of target microorganisms, even in mixed culture conditions, offering the following benefits:
- __Faster results__ compared to traditional methods: Some chromogenic media provide confirmed results within 24 hours.
- __Reliable visual detection__: Often, no further testing or subculture is required.
- __Multiple visual detection__: Distinguish multiple organisms on a single plate by their different colors.
- __Additional testing:__ Further testing is still possible directly from or on the media.
- __Combination with other biochemical tests__: This can include tests such as coagulase and lecithin tests.
- __Cost efficient__ and time saving solution.
[](https://www.sigmaaldrich.com)
* * *
## ChromoCult® Coliform Agar – A Culture Medium With 2 Different Chromogenic Substances
ChromoCult® Coliform Agar is a selective and differential chromogenic culture medium, suitable for microbiology laboratories that analyze food and water samples. This medium enables the detection, differentiation, and enumeration of *E. coli* and coliforms from drinking water and processed food matrices in under 24 hours. The careful selection of inhibitors in this selective media is crucial for the growth and recovery of damaged microorganisms. Notably, ChromoCult® Coliform Agar contains Tergitol® 7, which inhibits Gram-positive bacteria without adversely affecting the growth of the targeted coliforms or *E. coli*.
__Benefits of using ChromoCult® Coliform Agar:__
- __Time-saving:__ A fast method for simultaneous detection of coliforms and *E. coli* in water and processed food.
- __Safe analytical results:__ Reliable detection of injured bacteria due to carefully selected inhibitors, resulting in fewer chances of false
- __User-friendly:__ Clear detection of coliforms and *E. coli* even in the presence high amounts of accompanying flora. Simply add the recommend supplement
- __Cost-effective:__ Easy confirmation of *E. coli*; just add a drop of indole reagent to a colony, and the cherry color indicates whether it is *E. coli*.
- __Regulatory approval:__ Compliant to ISO 9308-1, approved by US-EPA, EU and AOAC™ as an alternative method for the detection of total coliforms and *E. coli* in water and processed food.
### ChromoCult® Coliform Agar ES (Enhanced Selectivity)[](https://www.sigmaaldrich.com)
Fresh food typically contains a high microbial load and generally does not include stressed or injured bacteria. The presence of accompanying flora requires a high selective culture medium to inhibit unwanted bacteria while promoting the growth of target organisms. To address this challenge, the well-established ChromoCult® Coliform Agar was modified. Tergitol® 7 was replaced with a combination of bile salts and propionate, which effectively inhibits accompanying bacteria.
[ChromoCult® Coliform Agar ES](https://www.sigmaaldrich.com/US/en/product/mm/100850) is a selective medium that allows for the simultaneous detection of coliforms and *E. coli* in samples with high bacterial bioburden. This makes it an ideal medium for detecting and counting total coliforms and *E. coli* in fresh foods as well as in water samples with higher bioburden. The combination of carefully selected peptones and the buffering capacity of MOPS creates optimal conditions for the rapid growth of coliforms and enhances the transformation of chromogenic substrates. The simultaneous detection of total coliforms and *E. coli* is achieved through the use of two specific chromogenic substrates.
[](https://www.sigmaaldrich.com)
[Shop All ChromoCult® Products](https://www.sigmaaldrich.com#shop-chromocult)
* * *
## ReadyCult® - A Culture Medium with A Combination of Fluorogenic and Chromogenic Substances[](https://www.sigmaaldrich.com)
ReadyCult® media enable detection of enterococci, coliforms, and *E. coli*. These media are supplied in a ready-to-use snap-pack format for fast and easy preparation. Results are obtained within 18 to 24 hours of incubation. Any color change of the broth to blue-green, even if only in the upper section, is a positive result.
#### ReadyCult® Coliforms 50/100[](https://www.sigmaaldrich.com)
[ReadyCult® Coliforms](https://www.sigmaaldrich.com/US/en/products/101298) 50 (for 50 mL water test sample) and ReadyCult® Coliforms 100 (for 100 mL water test sample) provide a rapid presence / absence test for the simultaneous detection of total coliforms and *E. coli* in drinking water. ReadyCult® is supplied as a premeasured, single use, sterilized packet of media.
When coliforms are present in the water sample, their *β*-galactosidase enzymes cleave the colorless X-GAL, causing the culture medium to change color from yellow to blue green. If *E. coli* is present among the coliforms, their *β*-glucuronidase enzymes cleave the substrate MUG (4-methylumbelliferyl-β-D-glucuronide), producing blue fluorescence that is visible under UV light (366 nm). However, it is important to note that the MUG reaction is not entirely specific to *E. coli* – a few other bacterial species can also cleave MUG (mug positive), and certain strains of *E. coli* may be MUG-negative. In case of any doubts, ReadyCult® Coliforms enables in contrast to other similar product available on the market, to a very simple and effective confirmation of the presence of *E. coli*: of a biochemical test can be performed directly in the sample by adding KOVAC's indole reagent. This reagent reacts with indole to produce a red ring in the presence of *E. coli*, indicating its ability to generate indole from the amino acid tryptophan. A positive result provides 99% certainty of the presence of *E. coli* and eliminates the need for additional 24-48 hour confirmation steps.
#### ReadyCult® Enterococci 100[](https://www.sigmaaldrich.com)
[ReadyCult® Enterococci](https://www.sigmaaldrich.com/US/en/product/mm/101299) 100 is a selective enrichment broth for the detection of Enterococcus and group D Streptococcus species in bacteriological water examinations. The peptone mixture ensures rapid growth of enterococci, while sodium azide effectively inhibits accompanying flora, particularly Gram-negative bacteria. In a reaction that is stimulated by selected peptones, the chromogenic substrate X-GLU is cleaved by the enzyme *β*-D-Glucosidase, which is characteristic of enterococci. This results in a color change of the broth to an intensive blue-green.
[Shop All ReadyCult® Products](https://www.sigmaaldrich.com#shop-readycult)
* * *
## MilliChrome™ Chromogenic Culture Media – Our Newest Chromogenic Media[](https://www.sigmaaldrich.com)
The new MilliChrome™ chromogenic culture media offer a convenient and rapid method for detecting common microbial contaminants and pathogens. They are well-suited for testing target organisms in quality control or research laboratories, as well as in non-critical areas of manufacturing facilities. These culture media contain substrates that specific bacterial species, genus, or serotypes can convert into a chromogen, a substance that imparts a specific color. This reaction is performed by an enzyme unique to the target organism. Consequently, colonies of the target organism on the agar exhibit the color of the chromogen, allowing for easy identification by the naked eye, often without the need for further testing. The cells remain culturable in or on any suitable medium.
MilliChrome™ media is the latest addition to our portfolio of chromogenic culture media. They include a dehydrated base medium, agar, and the chromogenic substrate for the identification of the target organism. Necessary supplements, as well as optional supplements that further enhances specificity, should be added after autoclaving, once the medium has cooled to between 45 and 50 °C. The MilliChrome™ media are suitable for a wide range of applications, including research and hygiene monitoring. Although, MilliChrome™ media do not have specific approvals, some of them are also useful for quality control of food samples in accordance with the relevant ISO guidelines.
[Shop All MilliChrome™ Products](https://www.sigmaaldrich.com#shop-millichrome)
* * *
## Related Products[](https://www.sigmaaldrich.com)
## ChromoCult® Culture Media
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
## FluoroCult® Culture Media
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
## Readycult® Culture Media
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
## MilliChrome™ Culture Media
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Related Articles__
- [Cell Culture Contamination Troubleshooting](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/mammalian-cell-culture/cell-culture-troubleshooting-contamination)
- [Streptococci - Overview of Detection, Identification, Differentiation and Cultivation Techniques](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/mammalian-cell-culture/streptococci-overview-of-detection)
- [Introduction to Microbial Media](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/microbial-cell-culture/microbial-media)
- [Antimicrobial Peptides](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/bacteriology/antimicrobial-peptides)
- [Inhibition of Cell Wall Biosynthesis by Antibiotics](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/bacteriology/how-antibiotics-work)
- [*Lactobacilli* Basics, Testing & Identification](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/bacteriology/lactobacilli)
- [Media for Staphylococcus Aureus Detection](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/bacteriology/staphylococcus-aureus)
- [Differentiation of Escherichia coli from coliforms](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/differentiation-of-e-coli-coliforms)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Microbial Culture Media Raw Materials](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/microbial-culture-media-raw-materials)
- [Microbial Culture Media](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/microbial-culture-media)
- [Microbial Identification Tests](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/microbial-identification-tests)
- [Pathogen Screening Kits, Instruments & Disposables](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/pathogen-screening-kits-instuments-and-disposables)
- [Quality indicator testing devices](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/quality-indicator-testing-devices)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)