Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Petrochem DHA
Detailed hydrocarbon analysis (DHA) is performed on long non-polar columns. Elution occurs in order of lowest boiling point to highest. Figure 1 shows a chromatogram of a gasoline sample using a 100 m Petrocol® DH column. The long column length and 60 °C isothermal oven temperature allow the separation of many components in this complex sample. Compounds identified range from propane (peak 1) to 1-methylnaphthalene (peak 96). Retention index libraries exist for this column, adding to its usefulness.
Petrochem DHA
Figure 1. Detailed Hydrocarbon Analysis (DHA)
Materials
Categories
Related Links