[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Metabolomics Research](https://www.sigmaaldrich.com/US/en/applications/research-disease-areas/metabolomics-research)Cholesterol Biosynthesis
# Cholesterol Biosynthesis
Cholesterol levels in the body come from two sources, dietary intake and biosynthesis. The majority of cholesterol utilized by healthy adults is synthesized in the liver, which produces ~70% of the total daily cholesterol requirement (~1 gram). The other 30% comes from dietary intake.
Biosynthesis of cholesterol generally takes place in the endoplasmic reticulum of hepatic cells and begins with acetyl- CoA, which is mainly derived from an oxidation reaction in the mitochondria. However, acetyl-CoA can also be derived from the cytoplasmic oxidation of ethanol by acetyl-CoA synthetase. Acetyl-CoA and acetoacetyl-CoA are converted to 3-hydroxy- 3-methylglutaryl-CoA (HMG-CoA) by HMG-CoA synthase.
[Search Related Products](https://www.sigmaaldrich.com#products)
__Figure 1.__IUBMB-Nicholson Metabolic Pathways Chart.
HMG-CoA is then converted to mevalonate by HMG-CoA reductase (HMGR). This reaction is completed with the aid of NADPH, which is used as a cofactor for all reduction reactions throughout cholesterol synthesis. Mevalonate undergoes a series of phosphorylations and a decarboxylation yielding the isoprenoid, isopentenyl pyrophosphate (IPP). A series of condensing reactions occur, catalyzed by squalene synthase, leading to the production of squalene. From squalene, lanosterol, the first of the sterols is formed. The conversion of lanosterol to cholesterol requires 19 additional reaction steps.
The conversion of HMG-CoA to mevalonate by HMG-CoA reductase is the rate-limiting step of cholesterol biosynthesis and is under strict regulatory control (__Figure 1__). HMGR is the target of compounds that are effective in lowering serum cholesterol levels. Human HMG-CoA reductase consists of a single polypeptide chain of 888 amino acids. The amino-terminal residues are membrane bound and reside in the endoplasmic reticulum membrane, while the catalytic site of the protein resides in its cytoplasmic, soluble carboxy-terminal portion. A linker region connects the two portions of the protein.
__Figure 1.__HMG-CoA Reductase.
## 3-Hydroxy-3-methyl glutar yl-CoA reductase human
HMG-CoA Reductase ; HMGR Hydroxy-3-methylglutaryl-CoA reductase catalyzes the committed step in cholesterol biosynthesis.
Consists of a single polypeptide chain of 888 amino acids. The amino-terminal residues are membrane bound and reside in the endoplasmic reticulum membrane, while the catalytic site of the protein resides in its cytoplasmic, soluble C-terminal portion. A linker region connects the two portions of the protein.
Supplied as a solution containing 250 μg protein in 50 mM Tris pH 7.5, with 5 mM DTT, 1:200 Protease Inhibitor Cocktail (Catalog Number P8340) and 50% (w/v) glycerol.
≥90% (SDS-PAGE)
activity: 2-8 units/mg protein
One unit will convert 1.0 μmole of NADPH to NADP + per minute at 37 °C. store at: -70 °C
## Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Related Articles__
- [UHPLC Vitamin D Analysis on Supel™ Carbon LC Columns](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/low-pressure-liquid-chromatography/uhplc-analysis-vitamin-d2-d3-metabolites-epimers-supel-carbon-lc-columns)
- [Analysis: Activated Nucleotide Sugars by LC-MS/MS](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/small-molecule-hplc/analysis-of-nucleotide-activated-sugars)
- [Untargeted profiling of *Glycyrrhiza glabra* extract with 2D LC MS](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/small-molecule-hplc/untargeted-profiling-of-glycyrrhiza-glabra-extract-with-2d-lc-ms)
- [Mitochondrial Stress and ROS](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/imaging-analysis-and-live-cell-imaging/mitochondrial-stress-and-ros)
- [Neuropeptidases](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/enzyme-activity-assays/neuropeptidases)
- [Neurotensin Receptors](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/enzyme-activity-assays/neurotensin-receptors)
- [α2-Adrenoceptors](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/protein-expression/alpha2-adrenoceptors)
- [β-Adrenoceptors](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/protein-expression/beta-adrenoceptors)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Lipids and Lipid Derivatives for Research](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/biochemicals/lipids)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)