[Skip to Content](https://www.sigmaaldrich.com#main-content) [![MilliporeSigma](https://www.sigmaaldrich.com/static/logos/purple/millipore_sigma.svg)](https://www.sigmaaldrich.com/US/en) Products Cart0 USEN Products ProductsApplicationsServicesResourcesSupport [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/US/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/US/en)[Applications](https://www.sigmaaldrich.com/US/en/applications)[Chemistry & Synthesis](https://www.sigmaaldrich.com/US/en/applications/chemistry-and-synthesis)Reaction Design & Optimization # Reaction Design & Optimization ![A close-up view of a laboratory automation system with multiple pipette tips positioned above several glass vials containing red liquid. ](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/applications/chemistry-and-synthesis/reaction-optimization-vab.jpg "Reaction optimization") Chemical reaction design and optimization is vital in organic synthesis research. By altering the reaction parameters (catalyst, pH, solvent, temperature, or time), certain outputs (cost savings, purity, selectivity, or yield) can be achieved. In optimizing chemical reactions, flexibility, precision and reproducibility are required of the synthesis tools with which the experiments are carried out. In designing chemical reactions, focus is placed on building a synthetic pathway to a target molecule from commercially available starting materials. A “disconnected approach” is typically taken, where focus is placed on the construction of key bonds. The process is broken down into simple steps, working backwards from the target molecule rather than forwards from the starting material. While many chemists resort to their extensive reaction knowledge to make these synthetic routes, there are now many software tools, such as [SYNTHIA™](https://www.sigmaaldrich.com/US/en/services/software-and-digital-platforms/synthia-retrosynthesis-software), which allow users to easily analyze custom pathways for known and novel molecules against search criteria.    * * * ## Related Products Slide 1 of 7 1 of 2 [![KitAlysis™ High-Throughput Buchwald-Hartwig Amination Reaction Screening Kit-Pack of 4](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/427/276/7492bdaf-54aa-4874-aaf0-290df24aec8d/640/7492bdaf-54aa-4874-aaf0-290df24aec8d.jpg) \ Sigma-Aldrich \ KITALYSIS-CN \ KitAlysis™ High-Throughput Buchwald-Hartwig Amination Reaction Screening Kit-Pack of 4](https://www.sigmaaldrich.com/US/en/product/aldrich/kitalysiscn) Quick View [![KitAlysis™ High-Throughput Copper C-N Cross-Coupling Reaction Screening Kit-Pack of 2](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/427/276/7492bdaf-54aa-4874-aaf0-290df24aec8d/640/7492bdaf-54aa-4874-aaf0-290df24aec8d.jpg) \ Sigma-Aldrich \ KITALYSIS-CUCN \ KitAlysis™ High-Throughput Copper C-N Cross-Coupling Reaction Screening Kit-Pack of 2](https://www.sigmaaldrich.com/US/en/product/aldrich/kitalysiscucn) Quick View [![KitAlysis™ High-Throughput Medium (5, 6, 7) Ring Closing Metathesis Reaction Screening Kit - Pack of 2](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/377/927/68a7355d-62d6-4702-a0f1-d3a0c3707877/640/68a7355d-62d6-4702-a0f1-d3a0c3707877.jpg) \ Sigma-Aldrich \ KITALYSIS-RCM \ KitAlysis™ High-Throughput Medium (5, 6, 7) Ring Closing Metathesis Reaction Screening Kit - Pack of 2](https://www.sigmaaldrich.com/US/en/product/aldrich/kitalysisrcm) Quick View [![KitAlysis™ High-Throughput 24 Pd Precatalyst Reaction Screening Kit-Pack of 2](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/427/276/7492bdaf-54aa-4874-aaf0-290df24aec8d/640/7492bdaf-54aa-4874-aaf0-290df24aec8d.jpg) \ Sigma-Aldrich \ KITALYSIS-24PD \ KitAlysis™ High-Throughput 24 Pd Precatalyst Reaction Screening Kit-Pack of 2](https://www.sigmaaldrich.com/US/en/product/aldrich/kitalysis24pd) Quick View [![KitAlysis™ High-Throughput Suzuki-Miyaura Cross-Coupling Reaction Screening Kit - Pack of 4](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/427/276/7492bdaf-54aa-4874-aaf0-290df24aec8d/640/7492bdaf-54aa-4874-aaf0-290df24aec8d.jpg) \ Sigma-Aldrich \ KITALYSIS-SM \ KitAlysis™ High-Throughput Suzuki-Miyaura Cross-Coupling Reaction Screening Kit - Pack of 4](https://www.sigmaaldrich.com/US/en/product/aldrich/kitalysissm) Quick View [![KitAlysis™ High-Throughput Base Screening Kit - Pack of 2](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/377/927/68a7355d-62d6-4702-a0f1-d3a0c3707877/640/68a7355d-62d6-4702-a0f1-d3a0c3707877.jpg) \ Sigma-Aldrich \ KITALYSISBASE \ KitAlysis™ High-Throughput Base Screening Kit - Pack of 2](https://www.sigmaaldrich.com/US/en/product/aldrich/kitalysisbase) Quick View [![KitAlysis™ High-Throughput Miyaura Borylation Reaction Screening Kit-Pack of 2](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/427/276/7492bdaf-54aa-4874-aaf0-290df24aec8d/640/7492bdaf-54aa-4874-aaf0-290df24aec8d.jpg) \ Sigma-Aldrich \ KITALYSIS-BOR \ KitAlysis™ High-Throughput Miyaura Borylation Reaction Screening Kit-Pack of 2](https://www.sigmaaldrich.com/US/en/product/aldrich/kitalysisbor) Quick View * * * ## Featured Categories [![Buchwald precatalysts activation scheme](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/biochemistry/buchwald-precat-activ-l-large-vac.jpg)](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/buchwald-catalysts-and-ligands) [Buchwald Catalysts & Ligands](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/buchwald-catalysts-and-ligands) Buchwald Catalysts and Ligands enable versatile cross-coupling reactions for C-C, C-N, and other bond formations. [Shop Products](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/buchwald-catalysts-and-ligands) [![Sample transition metals (iron, silver, irridium, rhodium, palladium, & nickel) in transition metal catalysts.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/korea/images/edited-transition-metal-catalysts.jpg)](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/transition-metal-catalysts) [Transition Metal Catalysts](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/transition-metal-catalysts) Buchwald Catalysts and Ligands enable versatile cross-coupling reactions for C-C, C-N, and other bond formations. [Shop Products](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/transition-metal-catalysts) [![Photoreactor in organic synthesis lab for photoredox catalysis](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/catalysts/photoreactor-in-lab.jpg)](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/photocatalysts) [Photocatalysts](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/photocatalysts) Photocatalysis utilizes visible light to activate a chemical reaction. Our extensive portfolio of catalysts and photoreactors enable consistent reactions in photoredox catalysis. [Shop Products](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/photocatalysts) [![A graphic representation of the chemical element Phosphorus from the periodic table, showing its atomic number ‘15’, and its atomic weight ‘30.973762’. ](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/catalysts/phosphine-ligands.jpg)](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/phosphine-ligands) [Phosphine Ligands](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/phosphine-ligands) Explore a wide array of phosphine ligands for cross-coupling reactions and diverse applications. [Shop Products](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/phosphine-ligands) Overview Related Articles & Protocols Support Several experimental methodologies can be employed in reaction optimization. In a trial and error, or one variable at a time, approach, all experimental inputs are kept constant, except one, to record a certain output. A series of reactions are performed until an optimum is determined. Another variable is then chosen, and the process repeats itself until all inputs have been probed and a set of optimal inputs have been established. A multi-parameter, or “design of experiments”, approach varies factors simultaneously from their lowest to highest value to find optimal conditions more efficiently. The different combinations are executed in the same set of experiments. Additional experiments are run between low and high factors to determine intrinsic variability. The values can be represented in a cube to illustrate the relations between the factors and the responses. For this optimization process to be successful, attention must be paid to reproducibility by performing the reactions in a systematic manner and controlled framework. After a viable synthetic pathway to synthesize the target molecule is found countless additional hours are put in to optimize each chemical reaction to make the product better, faster, or more efficient. Utilizing chemical reaction design optimization can lead to scientific breakthroughs more rapidly. ![Reaction Optimization Table](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/applications/chemistry-and-synthesis/reaction-optimization-table.jpg "Reaction Optimization Table") __Figure 1.__Reaction Optimization Table [Shop All Products](https://www.sigmaaldrich.com/search/reaction-design-%26-optimization?focus=products&page=1&perpage=30&sort=relevance&term=Reaction%20Design%20%26%20Optimization&type=product) [![Document Search](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/ecommerce/document-search.png)](https://www.sigmaaldrich.com/documents-search) [Looking for More Specific Information?](https://www.sigmaaldrich.com/documents-search) Visit our document search for data sheets, certificates and technical documentation. [Find Documents](https://www.sigmaaldrich.com/documents-search) ## Related Articles - [Molecular Sieves](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/reaction-design-and-optimization/molecular-sieves) Molecular sieves selectively adsorb molecules, crucial for various industrial processes. - [Macmillan Imidazolidinone Organocatalysts](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/reaction-design-and-optimization/organocatalyst) Discover Professor David MacMillan's groundbreaking metal-free asymmetric catalysis using imidazolidinone-based organocatalysts for versatile transformations. - [N-Heterocyclic Carbene Ligands](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/carbene-ligands) A wide range of NHC ligands are commonly available which exhibit high activities. - [Scale-Up Guide: Suzuki-Miyaura Cross-Coupling Reaction](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/s-m-kitalysis-scale-up) All of the preformed catalysts used in the kit are air and moisture stable complexes in their commercially available form. - [KitAlysis™ High-Throughput Screening Platform](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/kitalysis) KitAlysis High-Throughput Screening Kits enable efficient identification and optimization of catalytic reaction conditions. - [See All (88)](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search) ## Related Protocols - [Cleaning Laboratory Glassware](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/chemistry-and-synthesis/reaction-design-and-optimization/cleaning-glassware) Proper laboratory glassware cleaning techniques are crucial for high-quality results in experiments and research. - [Bestmann-Ohira Reagent: Alkynes from Aldehydes](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/chemistry-and-synthesis/reaction-design-and-optimization/alkyne-synthesis-bestmann-ohira-reagent) Acetylene chemistry's versatile applications from organic synthesis to bioorganic chemistry demand efficient synthesis methods. - [SnAP Reagents](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/chemistry-and-synthesis/reaction-design-and-optimization/snap-reagents) SnAP Reagents facilitate synthesis of saturated N-heterocycles for diverse structures. - [Step-by-Step Guide for KitAlysis™ Suzuki-Miyaura Cross-Coupling Reaction Screening Kit](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/chemistry-and-synthesis/reaction-design-and-optimization/kitalysis-suzuki-miyaura-set-up) A step-by-step protocol guide for KitAlysis Suzuki-Miyaura Cross-Coupling Reaction Screening Kit. - [KitAlysis™ High-Throughput Copper C-N Cross-Coupling Reaction Screening Kit](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/chemistry-and-synthesis/reaction-design-and-optimization/kitalysis-cucn-setup) KitAlysis™ Cu C-N (Buchwald-Hartwig) cross-coupling high-throughput screening kit. Detailed Set-Up User Guide and a downloadable excel file for calculations. - [See All (8)](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search) ### Find More Articles and Protocols Enter Keywords Search ## How Can We Help In case of any questions, please submit a [customer support request](https://www.sigmaaldrich.com/US/en/support/customer-support) or talk to our customer service team: Email [custserv@sial.com](mailto:custserv@sial.com) or call +1 (800) 244-1173 ## Additional Support - [Chromatogram Search](https://www.sigmaaldrich.com/chromatogram-search) Use the Chromatogram Search to identify unknown compounds in your sample. - [Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps) Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science. - [Customer Support Request](https://www.sigmaaldrich.com/US/en/support/customer-support) Customer support including help with orders, products, accounts, and website technical issues. - [FAQ](https://maestro.my.site.com/knowledgeportal/s/) Explore our Frequently Asked Questions for answers to commonly asked questions about our products and services. * * * __Featured Articles__ [Molecular Sieves](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/reaction-design-and-optimization/molecular-sieves) Molecular sieves selectively adsorb molecules, crucial for various industrial processes. [Macmillan Imidazolidinone Organocatalysts](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/reaction-design-and-optimization/organocatalyst) Discover Professor David MacMillan's groundbreaking metal-free asymmetric catalysis using imidazolidinone-based organocatalysts for versatile transformations. [Cleaning Laboratory Glassware](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/chemistry-and-synthesis/reaction-design-and-optimization/cleaning-glassware) Proper laboratory glassware cleaning techniques are crucial for high-quality results in experiments and research. [Bestmann-Ohira Reagent: Alkynes from Aldehydes](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/chemistry-and-synthesis/reaction-design-and-optimization/alkyne-synthesis-bestmann-ohira-reagent) Acetylene chemistry's versatile applications from organic synthesis to bioorganic chemistry demand efficient synthesis methods. Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars) Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions) Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices) Social Media [![LinkedIn icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/linkedin.svg)](https://www.linkedin.com/company/merck-life-science) [![X icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/x-icon.svg)](https://x.com/Merck_lifesci) [![Facebook Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/facebook.svg)](https://www.facebook.com/Merck.lifescience) [![Instagram Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/instagram.svg)](https://www.instagram.com/mercklifescience/) [![YouTube Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/youtube.svg)](https://www.youtube.com/@MerckLifeScience/featured) MilliporeSigma __Research. Development. Production.__ We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production. [![Vibrant M string](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/vibrant-m-string-1-rrvcrp.svg)](https://www.sigmaaldrich.com/US/en) [Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco) © 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies. Reproduction of any materials from the site is strictly forbidden without permission. [Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)