Fibronectin Coating Protocol
Important points to remember before start
  1. Ensure the cells are healthy and in adequate numbers.
  2. Fibronectin products provided as solutions can be stored for at least a year at 2 - 8 °C if kept sterile.
  3. Reconstitute fibronectin products provided as lyophilized powders in suitable solvents. Dissolve for at least 30 minutes at 37°C. A small amount of undissolved material may remain. This will not affect product performance. Reconstituted fibronectin solution can be stored in working aliquots at -20°C or lower.
  4. Vortexing or excessive agitation of fibronectin solutions should be avoided.
  5. Fibronectin coated cultureware can be stored for 2-4 weeks at 2-8°C in a closed sterile container or sterile sealable bags.
  6. Surface coverage concentration may differ with the cells being cultured. Protocol optimization is recommended to determine actual coating concentration for your cells. Recommended concentrations are listed in Table 1.
  7. Do not over-dry culture plates/coverslips post coating as this affects the attachment of cells.
  8. All the steps in the protocols need to be performed in sterile conditions.
Fibronectin coating protocol for culture ware
  1. Dilute fibronectin to the desired concentration. Optimum conditions for attachment are dependent on cell type and application. The typical coating concentration is 1 – 5 ug/cm2.
  2. Coat culture surface with a minimal volume.
  3. Once the entire area is covered, remove excess solution. This can be used to coat another well/plate.
  4. Air dry at least for 45 minutes at room temperature. before introducing cells and medium.
Product No. Description Coating concentration Dilute in
10838039001 Fibronectin from human plasma 5 μg/cm2 Sterile water
FIBRP-RO Fibronectin (pure) 5 μg/cm2 Sterile water
F0556 Fibronectin solution human fibroblasts 1-5 μg/cm2 sterile HBSS
F0635 Fibronectin rat plasma 1-5 μg/cm2 Sterile water
F0895 Fibronectin human plasma 1-5 μg/cm2 Sterile balanced salt solution
F1056 Fibronectin human plasma 1-5 μg/cm2 Sterile water
F1141 Fibronectin bovine plasma 1-5 μg/cm2 Sterile water
F2006 Fibronectin human plasma 1-5 μg/cm2 Sterile water
F2518 Fibronectin human foreskin fibroblasts 1-5 μg/cm2 Sterile water
F4759 Fibronectin bovine plasma 1-5 μg/cm2 Sterile water
FC010 Human Plasma Fibronectin Purified Protein 2-10 μg/cm2 Sterile balanced salt solution
FC014 Fibronectin, Bovine 2-10 μg/cm2 sterile balanced salt solution
S5171 Superfibronectin from human plasma 1 μg/cm2 Sterile PBS
ECM001 Fibronectin human, recombinant, expressed in HEK 1-5 μg/cm2 Sterile water
Table 1: Fibronectins for cell attachment
Fibronectin Proteolytic Fragments
Fibronectin fragments are obtained from plasma fibronectin by using proteolytic enzymes. These fragments are useful tools for mapping regions within the fibronectin molecule, which are responsible for specific binding to collagen and for other biologically relevant functions.
Product No. Description Binding activity
F9911 Fibronectin Proteolytic Fragment from human plasma, 30kDa Heparin-binding fragment
F0162 Fibronectin Proteolytic Fragment from human plasma, 45kDa Gelatin-binding fragment
F0287 Fibronectin Proteolytic Fragment from human plasma, 70kDa Heparin- and gelatin-binding fragment
F3542 Fibronectin Fragment III1-C human Fibronectin-binding fragment (self-assembly)
F1903 Fibronectin 40 kDa α Chymotryptic Fragment (Heparin-binding region), Human purified Heparin-binding fragment
F1904 Fibronectin 120 kDa α Chymotryptic Fragment (Cell Attachment region), Human purified cell attachment region of fibronectin
Table 2: Fibronectin fragments for binding studies
Click for Fibronectin Mimetics
Explore our 3D cell culture tools
Troubleshoot your cell culture challenges
Click for consistency in cell culture
Frequently Asked Questions (FAQs)
  1. What is the purity of fibronectin products?
    We do not determine the purity of fibronectin solutions. An SDS PAGE gel is run and one major fibronectin band of 120kDa is compared to past lots to ensure consistent quality. All our fibronectin powders are ≥95% pure.
  2. What is the best solvent to dilute fibronectin?
    The solvents that are compatible to dilute fibronectins are provided in Table 1
  3. How long can fibronectin be stored?
    Fibronectin products provided as solutions can be stored for a year at 2 - 8°C provided they are sterile. Diluted, unused fibronectin can be kept at 2 - 8°C for up to 1 week. Fibronectin-coated plates can be stored at 2 - 8°C for up to 4 weeks provided they are sealed well to prevent contamination and/or drying up of fibronectin. Do not use the product if discoloration or cracks appear on the surface of the coated material.
  4. What is the optimum concentration of fibronectin for coating?
    The coating concentrations of our fibronectins are suggested in Table 1. However, optimizing the concentrations that work for your cells is recommended.
  5. What is the best ECM protein to study stem cell differentiation?
    While laminin and fibronectin are widely used ECM proteins, the one that your cell needs should be determined by test experiments. Stem cells express varied integrins on the surface during different stages of differentiation. The choice of ECM protein would depend on the stage of differentiation you are interested in studying.
For additional questions contact our technical services here.