[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Western Blotting](https://www.sigmaaldrich.com/US/en/applications/protein-biology/western-blotting)Gelatine
# Gelatine
## What is gelatin?
Gelatin is a heterogeneous mixture of water-soluble proteins of high average molecular weights, present in collagen. The proteins are extracted by boiling skin, tendons, ligaments, bones, etc. in water.1 Type A gelatin is derived from acid-cured tissue and Type B gelatin is derived from lime-cured tissue.2
## What is gelatin used for?
Application using gelatin includes coating cell culture plates to improve cell attachment for a variety of cell types, addition to PCR to help stabilize Taq DNA polymerase,3 and use as a blocking reagent in Western blotting, ELISA, and immunohistochemistry.4 In bacteriology, gelatin can be used as a component of culture media for species differentiation.5 Additionally, as a biocompatible polymer, gelatin has been used as a delivery vehicle for the release of bioactive molecules6 and in the generation of scaffolds for tissue engineering applications.7
Industrial application includes the use of gelatin as a stabilizer, thickener, and texturizer in foods and in the manufacture of rubber substitutes, adhesives, cements, lithographic and printing inks, plastic compounds, artificial silk, photographic plates and films, matches, and light filters for mercury lamps.1 In the pharmaceutical industry, gelatin is used as a suspending agent, encapsulating agent, and tablet binder; and in veterinary applications, it is used as a plasma expander and hemostatic sponge.1
### Different Types of Gelatin
NameSourceStorage TempTarget Cells For AttachmentConcentration For UseCat. No.
Gelatinsolution2-8°CUsed for attachment of a variety of cell types100 ‑ 200 μg/cm2[G1393](https://www.sigmaaldrich.com/US/en/product/sigma/G1393)
Gelatin, powderfrom bovine skinRoom tempUsed for attachment of a variety of cell types100 ‑ 200 μg/cm2[G9391](https://www.sigmaaldrich.com/US/en/product/sigma/G9391)
Gelatin, powderfrom porcine skinRoom tempUsed for attachment of a variety of cell types100 ‑ 200 μg/cm2[G1890](https://www.sigmaaldrich.com/US/en/product/sigma/G1890)
Gelatin, lyophilized powderfrom porcine skinRoom tempUsed for attachment of a variety of cell types100 ‑ 200 μg/cm2[G9136](https://www.sigmaaldrich.com/US/en/product/sigma/G9136)
Gelatin, powderfrom cold water fish skin
Room temp
Used for attachment of a variety of cell types100 ‑ 200 μg/cm2[G7041](https://www.sigmaaldrich.com/catalog/product/sigma/g7041)
### References
1\.
Budavari S. 1996. Merck KGaA, Darmstadt, Germany Index. 742. 12th Ed:
2\.
Dieffenbach C, Dveksler GS. 1995. PCR Primer: A Laboratory Manual. NY: Cold Spring Harbor.
3\.
Vogt RV, Phillips DL, Omar Henderson L, Whitfield W, Spierto FW. 1987. Quantitative differences among various proteins as blocking agents for ELISA microtiter plates. Journal of Immunological Methods. 101(1):43-50. [https://doi.org/10.1016/0022-1759(87)90214-6](https://doi.org/10.1016/0022-1759%2887%2990214-6)
4\.
Levine M, Carpenter DC. 1923. GELATIN LIQUEFACTION BY BACTERIA. 8(4):297-306. [https://doi.org/10.1128/jb.8.4.297-306.1923](https://doi.org/10.1128/jb.8.4.297-306.1923)
5\.
Young S, Wong M, Tabata Y, Mikos AG. 2005. Gelatin as a delivery vehicle for the controlled release of bioactive molecules. Journal of Controlled Release. 109(1-3):256-274. [https://doi.org/10.1016/j.jconrel.2005.09.023](https://doi.org/10.1016/j.jconrel.2005.09.023)
6\.
Huang Y, Onyeri S, Siewe M, Moshfeghian A, Madihally SV. 2005. In vitro characterization of chitosan?gelatin scaffolds for tissue engineering. Biomaterials. 26(36):7616-7627. [https://doi.org/10.1016/j.biomaterials.2005.05.036](https://doi.org/10.1016/j.biomaterials.2005.05.036)
__Related Articles__
- [Improving Reproducibility: Best Practices for Antibodies](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/protein-biology/immunohistochemistry/improving-reproducibility-best-practices-for-antibodies)
- [Methods Used to Detect Proteins and Nucleic Acids Bound to Membranes](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/nucleic-acid-labeling-and-detection/detection-methods)
- [Antibody Standard Validation](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/elisa/antibody-standard-validation)
- [An Introduction to Antibodies: Antigens, Epitopes and Antibodies](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/elisa/antigens-epitopes-antibodies)
- [Antibody Generation | Monoclonal vs Polyclonal Antibodies](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/elisa/generation-of-antibodies)
- [Secondary Antibodies, Conjugates, and Kits](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/elisa/secondary-reagents)
- [Auto2D® Automated Gel Electrophoresis Device](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/gel-electrophoresis/automated-2d-gel-electrophoresis-device)
- [Protein Staining Reagents Selection Guide](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/gel-electrophoresis/electrophoresis-protein-staining-reagents)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.