[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Mammalian Cell Culture](https://www.sigmaaldrich.com/US/en/applications/cell-culture-and-cell-culture-analysis/cell-culture-by-cell-type/mammalian-cell-culture)Hematopoietic Cytokines
# Hematopoietic Cytokines
Hematopoietic cytokines are large family of extracellular ligands that stimulate hematopoietic cells to differentiate into eight principle types of blood cells. Numerous cytokines are involved in the regulation of hematopoiesis within a complex network of positive and negative regulators. Some cytokines have very narrow lineage specificities of their actions, while many others have rather broad and overlapping specificity ranges.1
Listed within this section are the cytokines whose predominant action appears to be the stimulation or regulation of hematopoietic cells. This includes GM-CSF, G-CSF, M-CSF, interleukins, EPO and TPO. There are a number of other cytokines that exert profound effects on the formation and maturation of hematopoietic cells, which include stem cell factor (SCF), flt-3/flk-2 ligand (FL) and [leukemia inhibitory factor (LIF)](https://www.sigmaaldrich.com/US/en/product/sigma/srp9001). Other cytokines or ligands such as jagged-1, transforming growth factor-β (TGF-β) and tumor necrosis factor-α (TNF-α) also play significant roles in modulating hematopoiesis.
| | |
|------------------------------------------------------------------------------------|------------------------------------------------------------------------------------|
| Cytokine | Function in hematopoiesis |
| [Erythropoietin (EPO)](https://www.sigmaaldrich.com/US/en/product/sigma/e5627) | Red blood cell production |
| [Flt-3](https://www.sigmaaldrich.com/US/en/product/sigma/srp6163) | Stimulation of stem and developing dendritic cells |
| Granulocyte-macrophage colony stimulating factor (GM-CSF) | Stimulation of diverse set of granulocyte-macrophage colonies |
| Granulocyte-colony stimulating factor (G-CSF) | granulocytic colony stimulation |
| Interleukin-2 (IL-2) | T-cell proliferation |
| Interleukin-3 (IL-3) | Granulocyte, macrophage, eosinophil, megakaryocyte and erythroid colony formation |
| Interleukin-5 (IL-5) | B-cell differentiation and eosinophil regulation |
| Interleukin-6 (IL-6) | B-cell differentiation |
| Interleukin-7 (IL-7) | T-lymphocyte induction |
| Interleukin-11 (IL-11) | Stimulation of megakaryocytes and plasmacytoma cell lines |
| Leukemia inhibitory factor (LIF) | Differentiation and suppression of clonogenicity of leukemic cells |
| Macrophage-colony stimulating factor (M-CSF) | Macrophage colony stimulation |
| [Stem cell factor (SCF)](https://www.sigmaaldrich.com/US/en/product/sigma/srp3234) | Proliferation of mast cells and stem cells |
| Thrombopoietin (TPO) | Regulation of platelet production; stimulation of megakaryocytes with IL-3 and SCF |
Table 1Regulation of Hematopoiesis by Cytokines
Two models were proposed to define the role of growth factors in hematopoietic differentiation.
- Instructive model assigns a direct role to cytokines in cell differentiation; cell fate is determined predominantly by the type of growth factor acting on the cell2
- Stochastic model suggests a pre-determined program for cell differentiation; growth factors are required specifically for survival and proliferation of committed progenitor cells3
Both models are based on evidence that growth factors control survival and proliferation of hematopoietic lineages and also transduce a genuine lineage-determining signal in hematopoiesis.1 The future studies focusing on elucidation of the unique role of growth factors in a particular situation and/or microenvironment in various disease situations they may prove to be of critical clinical value.
__Figure 1.__Hematopoietic cytokines stimulate hematopoietic cells to differentiate into principle types of blood cells.
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Adapted from Hematopoietic Cytokines by Jennifer Fries, BioFiles 2009, 4.5, 8.
### References
1\.
Metcalf D. 2008. Hematopoietic cytokines. 111(2):485-491. [https://doi.org/10.1182/blood-2007-03-079681](https://doi.org/10.1182/blood-2007-03-079681)
2\.
Borzillo GV, Ashmun RA, Sherr CJ. 1990. Macrophage lineage switching of murine early pre-B lymphoid cells expressing transduced fms genes.. Mol. Cell. Biol.. 10(6):2703-2714. [https://doi.org/10.1128/mcb.10.6.2703](https://doi.org/10.1128/mcb.10.6.2703)
3\.
Till JE, McCulloch EA, Siminovitch L. 1964. A STOCHASTIC MODEL OF STEM CELL PROLIFERATION, BASED ON THE GROWTH OF SPLEEN COLONY-FORMING CELLS. Proceedings of the National Academy of Sciences. 51(1):29-36. [https://doi.org/10.1073/pnas.51.1.29](https://doi.org/10.1073/pnas.51.1.29)
__Related Articles__
- [Extracellular Matrix Proteins and Tools for Cell Culture Optimization](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/3d-cell-culture/attachment-factors-for-cell-culture)
- [Human Serum Albumin (HSA)](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-culture-media-preparation/human-serum-albumin)
- [β-Alanine in Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-growth-and-maintenance/beta-alanine-cell-culture)
- [L-Asparagine in Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-growth-and-maintenance/l-asparagine-in-cell-culture)
- [L-Ornithine in Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-growth-and-maintenance/l-ornithine-cell-culture)
- [L-Tyrosine in Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-growth-and-maintenance/l-tyrosine-in-cell-culture)
- [Corning® High Content Screening Microplates – Frequently Asked Questions](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/imaging-analysis-and-live-cell-imaging/corning-high-screening-microplates)
- [Using Immortalized 16HBE14o- Human Bronchial Epithelial Cell Lines to Model Respiratory Lung Diseases](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/mammalian-cell-culture/16hbe-lung-epithelial-cell-lines)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Growth Factors and Cytokines](https://www.sigmaaldrich.com/US/en/products/cell-culture-and-analysis/cell-culture-supplements-and-reagents/growth-factors-and-cytokines)
- [Environmental Accounting Supplier Toolkit](https://www.sigmaaldrich.com/US/en/life-science/ssbi/sustainability-toolkits/environmental-accounting-supplier-toolkit)
- [Renewable Electricity Toolkit](https://www.sigmaaldrich.com/US/en/life-science/ssbi/sustainability-toolkits/renewable-electricity-toolkit)
- [Crown Ethers and Cryptands for Advanced Applications](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/crown-ethers-cryptands-advanced-applications)
- [Purification of Extracellular Vesicles with Eshmuno® Q Resin and Natrix® Q Membrane](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/membrane-chromatography/purification-extracellular-vesicles)
- [Sterilizing filters for final sterile filtration](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/pharma-and-biopharma-manufacturing/final-sterile-filtration-and-filling/sterilizing-grade-membrane-filters)
- [Single-use assemblies for final sterile filtration](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/pharma-and-biopharma-manufacturing/mobius-single-use-assemblies/single-use-filtration-assemblies)
- [Membrane Prefilters Improve Sterilizing Filter Capacity and Process Economics in Bioprocessing](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/prefilters-bioburden-control/membrane-prefiltration-improves-sterilizing-filter-capacity-and-process-economics)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.