Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Hepatocyte Growth Factor (HGF)
By: Jennifer Fries, BioFiles 2009, 4.5, 12.
Hepatocyte growth factor (HGF) is a potent mitogen for hepatocytes and a variety of other cells, including endothelial and epithelial cells, melanocytes, and keratinocytes. It also mediates epithelial morphogenesis. HGF is the ligand for a receptor encoded by the c-met proto-oncogene. c-Met expression has been observed in tumors from many sources, including stomach, lung, breast, kidney colon, liver, and melanoma, and is associated with tumor progression and metastasis.
back to top
Hepatocyte Growth Factor (HGF)
Product Description Source Recombinant Host Physical Form Assay Cell Type Affected Cat. No.
Hepatocyte Growth Factor human Baculovirus infected High-5 cells lyophilized powder The biological activity is measured by its ability to stimulate 3H-thymidine incorporation in the HGF responsive monkey epithelial cell line, 4MBr-5. renal tubular epithelial cell hepatocyte epidermal keratinocyte epidermal keratinocyte H1404-5UG
Hepatocyte Growth Factor human mouse NSO cells lyophilized powder The biological activity is measured by its ability to stimulate 3H-thymidine incorporation in the HGF responsive monkey epithelial cell line, 4MBr-5. hepatocyte epidermal keratinocyte renal tubular epithelial cell epidermal melanocyte H9661-5UG
back to top
Materials
References
  1. Birchmeier, C., et al., Met, metastasis, motility, and more. Nat. Rev. Mol. Cell Biol. 4, 915-925 (2003).
  2. Ma, P.C., et al., c-Met: structure, functions and potential for therapeutic inihibition. Cancer Metastasis Rev. 22, 309-325 (2003).
  3. Zhang, Y.W., and Vande Woude, G.F., HGF/SF-met signaling in the control of branching morphogensis and invasion. J. Cell Biochem. 88, 408-417 (2003).