[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[3D Cell Culture](https://www.sigmaaldrich.com/US/en/applications/cell-culture-and-cell-culture-analysis/cell-culture-by-technique/3d-cell-culture)Poly-L-Lysine Cell Attachment Protocol
# Poly-L-Lysine Cell Attachment Protocol
## Attach Cells to a Surface using Poly-lysine
Poly-L-lysine is a nonspecific attachment factor for cells and is useful in promoting cell adhesion to solid substrates by enhancing electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. After absorption to the culture surface, poly-lysine increases the number of positively charged cell binding sites. The following protocol applies to product numbers P4707\*, P4832\*, P7280, P6407, P7405, P9155, P6282, and P5899.
Optimal conditions must be determined for each cell line and application.
1. Add 50 mL of sterile tissue culture grade water to 5 mg of poly-lysine.
2. Aseptically coat culture surface with 1.0 mL/25 cm2 (only). Rock gently to ensure even coating of the culture surface.
3. After 5 minutes, remove solution by aspiration and thoroughly rinse surface with sterile tissue culture grade water.
4. Allow to dry at least 2 hours before introducing cells and medium.
\*__NOTE__: Step 1 is not necessary for poly-lysine solutions (P4707 and P4832).
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Related Articles__
- [Poly-lysine Types for Biomedical Applications](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-growth-and-maintenance/poly-lysine)
- [Cultrex ® 3-D Culture Matrix™ Laminin I Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/3d-cell-culture/3d-culture-matrix-laminin-i)
- [Cultrex® 3-D Culture Matrix™ Rat Collagen I Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/3d-cell-culture/3d-culture-matrix-rat-collagen-i)
- [Cultrex® 3D Spheroid BME Invasion Assay Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/3d-cell-culture/96-well-3d-spheroid-bme-cell-invasion-assay)
- [Cancer Stem Cell Tumorsphere Formation Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/3d-cell-culture/cancer-stem-cell-tumorsphere-formation)
- [Co-Culture of MCF-7 Cells and Human Primary Dermal Fibroblasts](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/3d-cell-culture/co-culture-mcf-7-cells-human-primary-dermal-fibroblasts)
- [Corning® Matrigel® Basement Membrane Matrix for 3D Culture *In Vitro*](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/3d-cell-culture/corning-matrigel-basement-membrane-matrix-for-3d-culture-in-vitro)
- [Corning® Spheroid Microplates](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/3d-cell-culture/corning-spheroid-microplates)
__Related Product Categories__
- [Attachment Factors](https://www.sigmaaldrich.com/US/en/products/cell-culture-and-analysis/cell-culture-supplements-and-reagents/attachment-factors)
- [Allergenic Fragrance Testing – Certified Materials](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/calibration-qualification-and-validation/allergenic-fragrance-testing)
- [Analysis of Alkylphenols Using New Internal Standards](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/calibration-qualification-and-validation/analysis-of-alkylphenols)
- [FAME Standard for Optimizing GC System Performance](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/calibration-qualification-and-validation/fame-standard-for)
- [Fast and Reliable Environmental Analysis of Aldehyde and Ketone Air Pollutants](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/calibration-qualification-and-validation/fast-and-reliable)
- [Hydrocarbon Oil Index in Samples](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/calibration-qualification-and-validation/hydrocarbon-oil-index)
- [Neonicotinoids](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/calibration-qualification-and-validation/neonicotinoids-bee-pesticides)
- [New GC-MS Method for Mycotoxin Analysis](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/calibration-qualification-and-validation/new-gc-ms-method-for-mycotoxin-analysis)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.