Atmosphere to Pascal Conversion
What is an Atmosphere (atm)
An atmosphere (atm) is a unit of measure for pressure defined as 101,325 pascals (Pa) or 101.325 kilopascals (kPa).
An atm is equivalent to the pressure exerted by the weight of approximately 1 cm2 (one square centimeter) of air.
1 atm = 101,325 Pa
1 Pa = 0.0000098692 atm
atm to Pa Conversion Formula
To convert atmospheres to pascals (atm to Pa) use the following formula:
Pa = atm/0.0000098692
Example: How many Pa is 10 atm?
Pa = 10 atm/0.0000098692
Pa = 1,013,250
10 atm = 1,013,250 Pa
Pa to atm Conversion
The pascal is an SI (international system) unit of measurement for pressure. To convert pascals to atmospheres (Pa to atm), simply reverse the formula.
atm = Pa/101,325
Example: How many atm is 10 pascals?
atm = 10/101,325
10 Pa = 0.000098692327 atm
Atmospheric Pressure (atm) to Pascal (Pa) Convertor
Use the atm to pa and pa to arm calculator to perform conversions. Enter in atm or Pa  in the box below to convert.
Pascals (pa)  
Atmosphere (atm)  
Atm to Pa Conversion Table
Use the following atm to Pa conversion table to easily find and convert atmospheres to pascals. Click for the downloadable
atm to Pa conversion table
.
Atmosphere (atm) Pascal (pa) Atmosphere (atm) Pascal (pa)
1 101325 51 5167575
2 202650 52 5268900
3 303975 53 5370225
4 405300 54 5471550
5 506625 55 5572875
6 607950 56 5674200
7 709275 57 5775525
8 810600 58 5876850
9 911925 59 5978175
10 1013250 60 6079500
11 1114575 61 6180825
12 1215900 62 6282150
13 1317225 63 6383475
14 1418550 64 6484800
15 1519875 65 6586125
16 1621200 66 6687450
17 1722525 67 6788775
18 1823850 68 6890100
19 1925175 69 6991425
20 2026500 70 7092750
21 2127825 71 7194075
22 2229150 72 7295400
23 2330475 73 7396725
24 2431800 74 7498050
25 2533125 75 7599375
26 2634450 76 7700700
27 2735775 77 7802025
28 2837100 78 7903350
29 2938425 79 8004675
30 3039750 80 8106000
31 3141075 81 8207325
32 3242400 82 8308650
33 3343725 83 8409975
34 3445050 84 8511300
35 3546375 85 8612625
36 3647700 86 8713950
37 3749025 87 8815275
38 3850350 88 8916600
39 3951675 89 9017925
40 4053000 90 9119250
41 4154325 91 9220575
42 4255650 92 9321900
43 4356975 93 9423225
44 4458300 94 9524550
45 4559625 95 9625875
46 4660950 96 9727200
47 4762275 97 9828525
48 4863600 98 9929850
49 4964925 99 10031175
50 5066250 100 10132500