Biomatrica Selection Guide
Biomatrica products enable researchers in academia, pharmaceutical drug development, and molecular diagnostics to stabilize ship and store biological samples for long periods of time with complete and rapid sample recovery, all at very affordable costs. Its SampleMatrix® proprietary core technology stabilizes and protects biological materials at room temperature without degradation. For help choosing the right product for your application, use the chart below. Application data is available on the product pages.
Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product was Designed for Energy Efficiency. To see how Stanford incorporated Biomatrica Technology and saved substantial amounts of energy, see their published study:
Biomatrica technology enables safe storage of biological material at room temperature. (PDF)
  Stability Products Pre-analytic Products
Product →
Application ↓
DNAstable Plus
DNAstable Blood
RNAstable LD
DNAgard Blood
DNAgard Tissue
Shipping & storage at RT X X
Forensics (storing extracts) X              
Forensics (concentrating extracts) X              
Controls for diagnostic and service labs   X   X X      
Biobanking   X X X X      
Sample concentration   X   X        
Sample collection at diagnostic and service labs             X  
Animal genotyping               X
Related Links