Dispensette® S and Dispensette® S Organic Chemical Compatibility Chart
The chart below lists chemical compatibility for Dispensette® S and Dispensette® S Organic from Brand. The recommendations reflect testing completed prior to publication. Always follow instructions in the operating manual of the instrument as well as the reagent manufacturer‘s specifications. In addition to these chemicals, a variety of organic and inorganic saline solutions (e.g., biological buffers), biological detergents and media for cell culture can be dispensed. Should you require information on chemicals not listed, please feel free to contact technical service.
Note: For dispensing HF, we recommend the use of the Dispensette® S Trace Analysis bottletop dispenser with platinum-iridium valve spring.
Reagent Dispensette S Dispensette S Organic
Acetaldehyde + +
Acetic acid (glacial), 100% + +
Acetic acid, ≤ 96% + +
Acetic anhydride   +
Acetone + +
Acetonitrile + +
Acetophenone   +
Acetyl chloride   +
Acetylacetone + +
Acrylic acid + +
Acrylonitrile + +
Adipic acid +  
Allyl alcohol + +
Aluminium chloride +  
Amino acids +  
Ammonia, ≤ 20% + +
Ammonia, 20-30%   +
Ammonium chloride +  
Ammonium fluoride +  
Ammonium sulfate +  
n-Amyl acetate + +
Amyl alcohol (Pentanol) + +
Amyl chloride (Chloropentane)   +
Aniline + +
Barium chloride +  
Benzaldehyde + +
Benzene (Benzol) + +
Benzine (Petroleum benzin), bp 70-180 °C   +
Benzoyl chloride + +
Benzyl alcohol + +
Benzylamine + +
Benzylchloride + +
Boric acid, ≤ 10% + +
Bromobenzene + +
Bromonaphthalene + +
Butanediol + +
1-Butanol + +
n-Butyl acetate + +
Butyl methyl ether + +
Butylamine + +
Butyric acid + +
Calcium carbonate +  
Calcium chloride +  
Calcium hydroxide +  
Calcium hypochlorite +  
Carbon tetrachloride   +
Chloro naphthalene + +
Chloroacetaldehyde, ≤ 45% + +
Chloroacetic acid + +
Chloroacetone + +
Chlorobenzene + +
Chlorobutane + +
Chloroform   +
Chlorosulfonic acid   +
Chromic acid, ≤ 50% + +
Chromosulfuric acid +  
Copper sulfate +  
Cresol   +
Cumene (Isopropyle benzene) + +
Cyclohexane   +
Cyclohexanone + +
Cyclopentane   +
Decane + +
1-Decanol + +
Dibenzyl ether + +
Dichloroacetic acid   +
Dichlorobenzene + +
Dichloroethane   +
Dichloroethylene   +
Dichloromethane   +
Diesel oil (Heating oil), bp 250-350 °C   +
Diethanolamine + +
Diethyl ether   +
Diethylamine + +
1.2 Diethylbenzene + +
Diethylene glycol + +
Dimethyl sulfoxide (DMSO) + +
Dimethylaniline +  
Dimethylformamide (DMF) + +
1.4 Dioxane   +
Diphenyl ether + +
Essential oil   +
Ethanol + +
Ethanolamine + +
Ethyl acetate + +
Ethylbenzene   +
Ethylene chloride   +
Fluoroacetic acid   +
Formaldehyde, ≤ 40% +  
Formamide + +
Formic acid, ≤ 100%   +
Glycerol + +
Glycol (Ethylene glycol) + +
Glycolic acid, ≤ 50% +  
Heating oil (Diesel oil), bp 250-350 °C    
Heptane   +
Hexane   +
Hexanoic acid + +
Hexanol + +
Hydriodic acid, ≤ 57% ** + +
Hydrobromic acid + +
Hydrochloric acid, ≤ 20% + +
Hydrochloric acid, 20-37% **   +
Hydrogen peroxide, ≤ 35%   +
Isoamyl alcohol + +
Isobutanol + +
Isooctane   +
Isopropanol (2-Propanol) + +
Isopropyl ether + +
Lactic acid +  
Methanol + +
Methoxybenzene + +
Methyl benzoate + +
Methyl butyl ether + +
Methyl ethyl ketone + +
Methyl formate + +
Methyl propyl ketone + +
Methylene chloride   +
Mineral oil (Engine oil) + +
Monochloroacetic acid + +
Nitric acid, ≤ 30% + +
Nitric acid, 30-70% */ **   +
Nitrobenzene + +
Oleic acid + +
Oxalic acid +  
n-Pentane   +
Peracetic acid   +
Perchloric acid + +
Perchloroethylene   +
Petroleum ether, bp 40-70 °C   +
Petroleum, bp 180-220 °C   +
Phenol + +
Phenylethanol + +
Phenylhydrazine + +
Phosphoric acid, ≤ 85% + +
Phosphoric acid, 85% + Sulfuric acid, 98%, 1:1 + +
Piperidine + +
Potassium chloride +  
Potassium dichromate +  
Potassium hydroxide +  
Potassium permanganate +  
Propionic acid + +
Propylene glycol (Propanediol) + +
Pyridine + +
Pyruvic acid + +
Salicylaldehyde + +
Scintilation fluid + +
Silver acetate +  
Silver nitrate +  
Sodium acetate +  
Sodium chloride +  
Sodium dichromate +  
Sodium fluoride +  
Sodium hydroxide, ≤ 30% +  
Sodium hypochlorite +  
Sulfuric acid, ≤ 98% + +
Tartaric acid +  
Tetrachloroethylene   +
Tetrahydrofuran (THF) * / **   +
Tetramethylammonium hydroxide +  
Toluene   +
Trichloroacetic acid   +
Trichlorobenzene   +
Trichloroethane   +
Trichloroethylene   +
Trichlorotrifluoro ethane   +
Triethanolamine + +
Triethylene glycol + +
Trifluoro ethane   +
Trifluoroacetic acid (TFA)   +
Turpentine   +
Urea +  
Xylene   +
Zinc chloride, ≤ 10% +  
Zinc sulfate, ≤ 10% +  
* Choose ETFE/PTFE adapters, if required, ** use PTFE seal for valve block
Note: For dispensing HF, we recommend the use of the Dispensette® S Trace Analysis bottletop dispenser with platinum-iridium valve spring.
Materials